Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130536955-130537515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC646743
Entrez Gene ID
646743
Gene Name
uncharacterized LOC646743
Gene Aliases
-
UniGene
Hs.634887
Gene Chromosome Location
Chr.2: 130536570-130549915 on Build GRCh38
Transcripts
NR_033930.1 XM_017004735.1

Gene Symbol
POTEI
Entrez Gene ID
653269
Gene Name
POTE ankyrin domain family member I
Gene Aliases
POTE2beta
UniGene
Hs.558782
Gene Chromosome Location
Chr.2: 130462245-131626428 on Build GRCh38
Transcripts
NR_033930.1 XM_017004735.1

SNP Detail

SNP ID Chromosome Location
rs71422874 Chr.2: 130537204-130537204 on Build GRCh38
rs1057278231 Chr.2: 130537394-130537394 on Build GRCh38
rs949027981 Chr.2: 130537480-130537480 on Build GRCh38
rs867537424 Chr.2: 130537030-130537030 on Build GRCh38
rs569868229 Chr.2: 130537173-130537173 on Build GRCh38
rs917503185 Chr.2: 130537473-130537473 on Build GRCh38
rs9653205 Chr.2: 130536977-130536977 on Build GRCh38
rs530870595 Chr.2: 130537251-130537251 on Build GRCh38
rs934829976 Chr.2: 130537368-130537368 on Build GRCh38

View Full Product Details