Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23399625-23400200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC8P1
Entrez Gene ID
150244
Gene Name
zinc finger DHHC-type containing 8 pseudogene 1
Gene Aliases
ZDHHC8P
UniGene
Hs.386693
Gene Chromosome Location
Chr.22: 23390605-23402612 on Build GRCh38
Transcripts
NR_003950.1

SNP Detail

SNP ID Chromosome Location
rs180738580 Chr.22: 23399800-23399800 on Build GRCh38
rs13054327 Chr.22: 23400121-23400121 on Build GRCh38
rs148919568 Chr.22: 23399857-23399857 on Build GRCh38
rs979592895 Chr.22: 23399667-23399667 on Build GRCh38
rs915724687 Chr.22: 23400179-23400179 on Build GRCh38
rs375669030 Chr.22: 23400099-23400099 on Build GRCh38
rs1056989575 Chr.22: 23399709-23399709 on Build GRCh38
rs982441661 Chr.22: 23399935-23399935 on Build GRCh38
rs117039393 Chr.22: 23399662-23399662 on Build GRCh38
rs73880815 Chr.22: 23399801-23399801 on Build GRCh38
rs41277531 Chr.22: 23399691-23399691 on Build GRCh38
rs1015244943 Chr.22: 23400136-23400136 on Build GRCh38
rs112158649 Chr.22: 23399735-23399735 on Build GRCh38
rs903829158 Chr.22: 23400002-23400002 on Build GRCh38
rs369248510 Chr.22: 23399923-23399923 on Build GRCh38
rs780696755 Chr.22: 23399839-23399839 on Build GRCh38
rs1031137978 Chr.22: 23399748-23399748 on Build GRCh38
rs1012228460 Chr.22: 23399729-23399729 on Build GRCh38
rs768084972 Chr.22: 23399987-23399987 on Build GRCh38
rs779156363 Chr.22: 23399856-23399856 on Build GRCh38
rs374814932 Chr.22: 23400169-23400169 on Build GRCh38
rs112045685 Chr.22: 23399862-23399862 on Build GRCh38
rs376484775 Chr.22: 23399978-23399978 on Build GRCh38
rs546596946 Chr.22: 23399705-23399705 on Build GRCh38
rs189363237 Chr.22: 23399960-23399960 on Build GRCh38
rs538093774 Chr.22: 23399797-23399797 on Build GRCh38
rs117550380 Chr.22: 23399661-23399661 on Build GRCh38
rs13054421 Chr.22: 23399831-23399831 on Build GRCh38
rs181289871 Chr.22: 23400014-23400014 on Build GRCh38
rs547547045 Chr.22: 23400046-23400046 on Build GRCh38
rs540738252 Chr.22: 23399679-23399679 on Build GRCh38
rs956909304 Chr.22: 23399672-23399672 on Build GRCh38
rs531366772 Chr.22: 23399991-23399991 on Build GRCh38
rs749502418 Chr.22: 23399827-23399829 on Build GRCh38
rs774266293 Chr.22: 23399979-23399979 on Build GRCh38
rs1043387122 Chr.22: 23399996-23399997 on Build GRCh38
rs1008382226 Chr.22: 23399858-23399858 on Build GRCh38
rs376575012 Chr.22: 23400064-23400064 on Build GRCh38
rs369574846 Chr.22: 23400029-23400029 on Build GRCh38
rs371032015 Chr.22: 23400075-23400075 on Build GRCh38
rs750318232 Chr.22: 23399653-23399653 on Build GRCh38
rs368047578 Chr.22: 23399794-23399794 on Build GRCh38
rs919668135 Chr.22: 23399701-23399701 on Build GRCh38
rs1027636193 Chr.22: 23400109-23400109 on Build GRCh38
rs371419720 Chr.22: 23400146-23400146 on Build GRCh38
rs968150771 Chr.22: 23399738-23399738 on Build GRCh38
rs561478770 Chr.22: 23399665-23399665 on Build GRCh38
rs931096049 Chr.22: 23399702-23399702 on Build GRCh38
rs533887952 Chr.22: 23400022-23400022 on Build GRCh38
rs118023203 Chr.22: 23399977-23399977 on Build GRCh38
rs201942212 Chr.22: 23400159-23400160 on Build GRCh38
rs1024093955 Chr.22: 23399737-23399737 on Build GRCh38
rs553823666 Chr.22: 23399821-23399821 on Build GRCh38
rs372060930 Chr.22: 23399820-23399820 on Build GRCh38
rs900016131 Chr.22: 23400106-23400106 on Build GRCh38
rs924138751 Chr.22: 23399669-23399669 on Build GRCh38
rs561183672 Chr.22: 23399879-23399879 on Build GRCh38
rs369868374 Chr.22: 23399808-23399808 on Build GRCh38
rs538945223 Chr.22: 23399855-23399855 on Build GRCh38
rs776302072 Chr.22: 23399995-23399995 on Build GRCh38
rs372232071 Chr.22: 23399799-23399799 on Build GRCh38
rs375697656 Chr.22: 23400117-23400117 on Build GRCh38
rs2003752 Chr.22: 23399918-23399918 on Build GRCh38
rs560099103 Chr.22: 23399972-23399972 on Build GRCh38
rs1000960470 Chr.22: 23399742-23399742 on Build GRCh38
rs908353971 Chr.22: 23399807-23399807 on Build GRCh38
rs374858569 Chr.22: 23399788-23399788 on Build GRCh38
rs371490947 Chr.22: 23399787-23399787 on Build GRCh38
rs144302651 Chr.22: 23400013-23400013 on Build GRCh38
rs987024901 Chr.22: 23399781-23399781 on Build GRCh38
rs867010863 Chr.22: 23399814-23399814 on Build GRCh38
rs568936773 Chr.22: 23399791-23399791 on Build GRCh38
rs959745712 Chr.22: 23400143-23400143 on Build GRCh38
rs987054621 Chr.22: 23399678-23399678 on Build GRCh38
rs368575734 Chr.22: 23399764-23399764 on Build GRCh38
rs539465768 Chr.22: 23400116-23400116 on Build GRCh38
rs766469827 Chr.22: 23400087-23400087 on Build GRCh38
rs1002720789 Chr.22: 23399706-23399706 on Build GRCh38
rs552187661 Chr.22: 23399988-23399988 on Build GRCh38
rs535136074 Chr.22: 23399658-23399658 on Build GRCh38
rs544829753 Chr.22: 23399867-23399867 on Build GRCh38
rs41277533 Chr.22: 23399727-23399727 on Build GRCh38
rs942396818 Chr.22: 23399682-23399682 on Build GRCh38
rs992988373 Chr.22: 23400165-23400165 on Build GRCh38
rs113578719 Chr.22: 23399874-23399874 on Build GRCh38
rs976079362 Chr.22: 23399908-23399908 on Build GRCh38
rs562844448 Chr.22: 23399990-23399990 on Build GRCh38
rs761497023 Chr.22: 23400003-23400003 on Build GRCh38

View Full Product Details