Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15205554-15206100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A4P1
Entrez Gene ID
344875
Gene Name
collagen type VI alpha 4 pseudogene 1
Gene Aliases
COL6A4, COL6A4P, DIVA, DVWA, VWA6
UniGene
Hs.722756
Gene Chromosome Location
Chr.3: 15165362-15205959 on Build GRCh38
Transcripts
NR_027927.1

SNP Detail

SNP ID Chromosome Location
rs143705406 Chr.3: 15205780-15205780 on Build GRCh38
rs796726113 Chr.3: 15205779-15205779 on Build GRCh38
rs997706255 Chr.3: 15205728-15205728 on Build GRCh38
rs767106761 Chr.3: 15205657-15205657 on Build GRCh38
rs989986990 Chr.3: 15206060-15206060 on Build GRCh38
rs1006987471 Chr.3: 15205740-15205740 on Build GRCh38
rs989955938 Chr.3: 15205843-15205843 on Build GRCh38
rs113668391 Chr.3: 15205822-15205822 on Build GRCh38
rs927383682 Chr.3: 15205834-15205834 on Build GRCh38
rs997952054 Chr.3: 15205737-15205737 on Build GRCh38
rs914503058 Chr.3: 15205854-15205854 on Build GRCh38
rs191046180 Chr.3: 15205838-15205838 on Build GRCh38
rs945908814 Chr.3: 15205867-15205867 on Build GRCh38
rs1039374585 Chr.3: 15205635-15205635 on Build GRCh38
rs981943102 Chr.3: 15205873-15205873 on Build GRCh38
rs550823116 Chr.3: 15205900-15205900 on Build GRCh38
rs565080019 Chr.3: 15205869-15205869 on Build GRCh38
rs866126400 Chr.3: 15205891-15205891 on Build GRCh38
rs1034863311 Chr.3: 15206045-15206045 on Build GRCh38
rs754675527 Chr.3: 15206031-15206031 on Build GRCh38
rs1025882648 Chr.3: 15205975-15205975 on Build GRCh38
rs958746112 Chr.3: 15206059-15206059 on Build GRCh38
rs1033716967 Chr.3: 15205815-15205815 on Build GRCh38
rs532537207 Chr.3: 15206073-15206073 on Build GRCh38
rs901597255 Chr.3: 15205721-15205721 on Build GRCh38
rs1050616008 Chr.3: 15205949-15205949 on Build GRCh38
rs1029383604 Chr.3: 15205758-15205758 on Build GRCh38
rs986745540 Chr.3: 15206001-15206001 on Build GRCh38
rs58318356 Chr.3: 15205618-15205618 on Build GRCh38
rs750150231 Chr.3: 15205705-15205705 on Build GRCh38
rs889973364 Chr.3: 15205739-15205739 on Build GRCh38
rs753675630 Chr.3: 15206009-15206009 on Build GRCh38
rs945710502 Chr.3: 15205667-15205667 on Build GRCh38
rs957608370 Chr.3: 15205816-15205816 on Build GRCh38
rs375986796 Chr.3: 15205969-15205969 on Build GRCh38
rs577364018 Chr.3: 15205998-15205998 on Build GRCh38
rs967189690 Chr.3: 15206078-15206078 on Build GRCh38
rs546338323 Chr.3: 15205997-15205997 on Build GRCh38
rs755853102 Chr.3: 15205832-15205832 on Build GRCh38
rs148046701 Chr.3: 15205966-15205966 on Build GRCh38
rs772852337 Chr.3: 15205603-15205603 on Build GRCh38
rs941241230 Chr.3: 15205985-15205985 on Build GRCh38
rs888368460 Chr.3: 15205613-15205613 on Build GRCh38
rs530816922 Chr.3: 15205889-15205889 on Build GRCh38
rs899511833 Chr.3: 15206071-15206071 on Build GRCh38
rs888833649 Chr.3: 15205950-15205950 on Build GRCh38
rs914448534 Chr.3: 15206067-15206067 on Build GRCh38
rs540337697 Chr.3: 15205674-15205674 on Build GRCh38
rs182812029 Chr.3: 15205902-15205902 on Build GRCh38
rs994862023 Chr.3: 15205972-15205972 on Build GRCh38
rs143147141 Chr.3: 15205607-15205607 on Build GRCh38
rs899608254 Chr.3: 15205672-15205672 on Build GRCh38

View Full Product Details