Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141234692-141235248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB18P
Entrez Gene ID
54660
Gene Name
protocadherin beta 18 pseudogene
Gene Aliases
PCDH-psi2, PCDHB18
UniGene
Hs.676071
Gene Chromosome Location
Chr.5: 141234333-141237529 on Build GRCh38
Transcripts
NR_001281.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6033597 c.113G>A p.R38Q Chr.5: 141234826-141234826 on Build GRCh38
3702733 c.375C>T p.H125H Chr.5: 141235088-141235088 on Build GRCh38
5515981 c.470C>T p.T157M Chr.5: 141235183-141235183 on Build GRCh38
5953729 c.468A>G p.L156L Chr.5: 141235181-141235181 on Build GRCh38
4432104 c.22_23insG p.A10fs*6 Chr.5: 141234735-141234736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561851543 Chr.5: 141235222-141235222 on Build GRCh38
rs541962708 Chr.5: 141235203-141235203 on Build GRCh38
rs1009176780 Chr.5: 141234850-141234850 on Build GRCh38
rs145435798 Chr.5: 141234792-141234792 on Build GRCh38
rs1050172475 Chr.5: 141234813-141234813 on Build GRCh38
rs115807044 Chr.5: 141235156-141235156 on Build GRCh38
rs890277993 Chr.5: 141234825-141234825 on Build GRCh38
rs140239305 Chr.5: 141235035-141235035 on Build GRCh38
rs770639417 Chr.5: 141235137-141235137 on Build GRCh38
rs185594704 Chr.5: 141234761-141234761 on Build GRCh38
rs535554930 Chr.5: 141234843-141234843 on Build GRCh38
rs184425682 Chr.5: 141234735-141234735 on Build GRCh38
rs782532130 Chr.5: 141234822-141234822 on Build GRCh38
rs781811713 Chr.5: 141235223-141235223 on Build GRCh38
rs781988439 Chr.5: 141234895-141234895 on Build GRCh38
rs782819102 Chr.5: 141234927-141234927 on Build GRCh38
rs553007821 Chr.5: 141235150-141235150 on Build GRCh38
rs994763266 Chr.5: 141234942-141234942 on Build GRCh38
rs182253231 Chr.5: 141234754-141234754 on Build GRCh38
rs782297384 Chr.5: 141234905-141234905 on Build GRCh38
rs782105106 Chr.5: 141235065-141235065 on Build GRCh38
rs781919327 Chr.5: 141234902-141234902 on Build GRCh38
rs1050122138 Chr.5: 141235204-141235204 on Build GRCh38
rs782630005 Chr.5: 141234834-141234834 on Build GRCh38
rs781887208 Chr.5: 141235124-141235124 on Build GRCh38
rs567746958 Chr.5: 141234743-141234743 on Build GRCh38
rs782756865 Chr.5: 141235076-141235076 on Build GRCh38
rs181575572 Chr.5: 141235145-141235145 on Build GRCh38
rs61738060 Chr.5: 141234736-141234736 on Build GRCh38
rs782694623 Chr.5: 141234794-141234794 on Build GRCh38
rs535279098 Chr.5: 141235087-141235087 on Build GRCh38
rs190170963 Chr.5: 141234851-141234851 on Build GRCh38
rs931727084 Chr.5: 141234799-141234799 on Build GRCh38
rs550637181 Chr.5: 141234738-141234738 on Build GRCh38
rs781858573 Chr.5: 141235158-141235158 on Build GRCh38
rs782697364 Chr.5: 141234726-141234726 on Build GRCh38
rs61733834 Chr.5: 141235098-141235098 on Build GRCh38
rs566583324 Chr.5: 141234803-141234803 on Build GRCh38
rs781999319 Chr.5: 141234991-141234991 on Build GRCh38
rs569122528 Chr.5: 141234947-141234947 on Build GRCh38
rs782345729 Chr.5: 141234889-141234889 on Build GRCh38
rs782398685 Chr.5: 141234930-141234930 on Build GRCh38
rs781913768 Chr.5: 141235110-141235110 on Build GRCh38
rs781943268 Chr.5: 141235079-141235079 on Build GRCh38
rs782724765 Chr.5: 141234750-141234750 on Build GRCh38
rs782479097 Chr.5: 141234742-141234742 on Build GRCh38
rs782484397 Chr.5: 141235165-141235165 on Build GRCh38
rs112268051 Chr.5: 141234734-141234735 on Build GRCh38
rs372147512 Chr.5: 141234741-141234741 on Build GRCh38
rs969416383 Chr.5: 141235021-141235021 on Build GRCh38
rs541999317 Chr.5: 141234725-141234725 on Build GRCh38
rs1027648539 Chr.5: 141234989-141234989 on Build GRCh38
rs928112009 Chr.5: 141234737-141234737 on Build GRCh38
rs782279691 Chr.5: 141234795-141234795 on Build GRCh38
rs988690398 Chr.5: 141235112-141235112 on Build GRCh38

View Full Product Details