Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29751045-29751583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFITM4P
Entrez Gene ID
340198
Gene Name
interferon induced transmembrane protein 4 pseudogene
Gene Aliases
dJ377H14.5
UniGene
Hs.450189
Gene Chromosome Location
Chr.6: 29750807-29751148 on Build GRCh38
Transcripts
NR_001590.1

SNP Detail

SNP ID Chromosome Location
rs201826155 Chr.6: 29751470-29751470 on Build GRCh38
rs748748083 Chr.6: 29751231-29751231 on Build GRCh38
rs114038840 Chr.6: 29751413-29751413 on Build GRCh38
rs190022429 Chr.6: 29751483-29751483 on Build GRCh38
rs748135212 Chr.6: 29751166-29751166 on Build GRCh38
rs978751611 Chr.6: 29751425-29751425 on Build GRCh38
rs138248603 Chr.6: 29751464-29751464 on Build GRCh38
rs1009215012 Chr.6: 29751502-29751502 on Build GRCh38
rs549389829 Chr.6: 29751404-29751404 on Build GRCh38
rs1043769990 Chr.6: 29751544-29751544 on Build GRCh38
rs114556323 Chr.6: 29751319-29751319 on Build GRCh38
rs970059030 Chr.6: 29751177-29751177 on Build GRCh38
rs960146280 Chr.6: 29751122-29751122 on Build GRCh38
rs563952034 Chr.6: 29751320-29751320 on Build GRCh38
rs1022185580 Chr.6: 29751262-29751262 on Build GRCh38
rs565896490 Chr.6: 29751447-29751447 on Build GRCh38
rs1014319606 Chr.6: 29751359-29751359 on Build GRCh38
rs879476445 Chr.6: 29751450-29751451 on Build GRCh38
rs898848164 Chr.6: 29751495-29751495 on Build GRCh38
rs376207012 Chr.6: 29751543-29751543 on Build GRCh38
rs796700054 Chr.6: 29751418-29751418 on Build GRCh38
rs55884048 Chr.6: 29751178-29751178 on Build GRCh38
rs564496759 Chr.6: 29751280-29751280 on Build GRCh38
rs925987769 Chr.6: 29751428-29751428 on Build GRCh38
rs529660373 Chr.6: 29751403-29751403 on Build GRCh38
rs1045355237 Chr.6: 29751486-29751486 on Build GRCh38
rs772510273 Chr.6: 29751476-29751476 on Build GRCh38
rs922768948 Chr.6: 29751467-29751467 on Build GRCh38
rs56304249 Chr.6: 29751092-29751092 on Build GRCh38
rs533332566 Chr.6: 29751312-29751312 on Build GRCh38
rs142834053 Chr.6: 29751494-29751494 on Build GRCh38
rs181712476 Chr.6: 29751215-29751215 on Build GRCh38
rs903551324 Chr.6: 29751557-29751557 on Build GRCh38
rs892234899 Chr.6: 29751499-29751499 on Build GRCh38
rs926234270 Chr.6: 29751186-29751186 on Build GRCh38
rs186672680 Chr.6: 29751235-29751235 on Build GRCh38
rs754333158 Chr.6: 29751491-29751491 on Build GRCh38
rs201654301 Chr.6: 29751385-29751385 on Build GRCh38
rs777425445 Chr.6: 29751193-29751193 on Build GRCh38
rs571306198 Chr.6: 29751089-29751089 on Build GRCh38
rs1051475694 Chr.6: 29751234-29751234 on Build GRCh38
rs915900748 Chr.6: 29751175-29751175 on Build GRCh38
rs116165542 Chr.6: 29751106-29751106 on Build GRCh38
rs565015617 Chr.6: 29751304-29751304 on Build GRCh38
rs1037545824 Chr.6: 29751266-29751266 on Build GRCh38
rs761949829 Chr.6: 29751521-29751521 on Build GRCh38
rs558243288 Chr.6: 29751068-29751068 on Build GRCh38
rs542239870 Chr.6: 29751093-29751093 on Build GRCh38
rs992933899 Chr.6: 29751168-29751168 on Build GRCh38
rs1009246081 Chr.6: 29751256-29751256 on Build GRCh38
rs780823556 Chr.6: 29751134-29751134 on Build GRCh38
rs551555374 Chr.6: 29751465-29751465 on Build GRCh38
rs960173068 Chr.6: 29751347-29751347 on Build GRCh38
rs775677255 Chr.6: 29751484-29751484 on Build GRCh38
rs542191821 Chr.6: 29751202-29751202 on Build GRCh38
rs111324950 Chr.6: 29751255-29751255 on Build GRCh38
rs1007502915 Chr.6: 29751282-29751282 on Build GRCh38
rs761253189 Chr.6: 29751485-29751485 on Build GRCh38

View Full Product Details