Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53335080-53335645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL5
Entrez Gene ID
60481
Gene Name
ELOVL fatty acid elongase 5
Gene Aliases
HELO1, SCA38, dJ483K16.1
UniGene
Hs.520189
Gene Chromosome Location
Chr.6: 53267398-53349179 on Build GRCh38
Transcripts
NM_001242828.1 NM_001242830.1 NM_001242831.1 NM_001301856.1 NM_021814.4 NR_046241.1

Gene Symbol
RPS16P5
Entrez Gene ID
647190
Gene Name
ribosomal protein S16 pseudogene 5
Gene Aliases
RPS16_3_702
UniGene
Hs.520189
Gene Chromosome Location
Chr.6: 53334460-53337617 on Build GRCh38
Transcripts
NM_001242828.1 NM_001242830.1 NM_001242831.1 NM_001301856.1 NM_021814.4 NR_046241.1

SNP Detail

SNP ID Chromosome Location
rs73740717 Chr.6: 53335320-53335320 on Build GRCh38
rs1039069564 Chr.6: 53335127-53335127 on Build GRCh38
rs1044885671 Chr.6: 53335399-53335399 on Build GRCh38
rs370012480 Chr.6: 53335543-53335543 on Build GRCh38
rs749258890 Chr.6: 53335482-53335482 on Build GRCh38
rs149042354 Chr.6: 53335140-53335140 on Build GRCh38
rs999955944 Chr.6: 53335287-53335287 on Build GRCh38
rs911566151 Chr.6: 53335333-53335333 on Build GRCh38
rs570271753 Chr.6: 53335361-53335361 on Build GRCh38
rs890177284 Chr.6: 53335104-53335104 on Build GRCh38
rs568480748 Chr.6: 53335227-53335227 on Build GRCh38
rs1035437015 Chr.6: 53335237-53335237 on Build GRCh38
rs1056608470 Chr.6: 53335541-53335541 on Build GRCh38
rs934148 Chr.6: 53335522-53335522 on Build GRCh38
rs975543876 Chr.6: 53335580-53335580 on Build GRCh38
rs1012721092 Chr.6: 53335609-53335609 on Build GRCh38
rs143175632 Chr.6: 53335618-53335618 on Build GRCh38
rs768573575 Chr.6: 53335483-53335483 on Build GRCh38
rs746559267 Chr.6: 53335228-53335228 on Build GRCh38
rs964781230 Chr.6: 53335401-53335401 on Build GRCh38
rs534144651 Chr.6: 53335529-53335529 on Build GRCh38
rs895326693 Chr.6: 53335574-53335574 on Build GRCh38
rs527500264 Chr.6: 53335293-53335293 on Build GRCh38
rs771494824 Chr.6: 53335415-53335415 on Build GRCh38
rs75380947 Chr.6: 53335279-53335279 on Build GRCh38
rs758368735 Chr.6: 53335201-53335201 on Build GRCh38
rs911727449 Chr.6: 53335390-53335390 on Build GRCh38
rs780869450 Chr.6: 53335305-53335305 on Build GRCh38
rs188934598 Chr.6: 53335433-53335433 on Build GRCh38
rs921516922 Chr.6: 53335300-53335300 on Build GRCh38
rs376069868 Chr.6: 53335280-53335280 on Build GRCh38
rs184292294 Chr.6: 53335414-53335414 on Build GRCh38
rs921432986 Chr.6: 53335583-53335583 on Build GRCh38
rs2397142 Chr.6: 53335501-53335501 on Build GRCh38
rs1012336498 Chr.6: 53335254-53335254 on Build GRCh38
rs1003702554 Chr.6: 53335206-53335206 on Build GRCh38
rs761842258 Chr.6: 53335502-53335502 on Build GRCh38
rs956910422 Chr.6: 53335488-53335488 on Build GRCh38
rs564241061 Chr.6: 53335340-53335340 on Build GRCh38
rs753026567 Chr.6: 53335509-53335509 on Build GRCh38
rs940421739 Chr.6: 53335335-53335335 on Build GRCh38
rs988237014 Chr.6: 53335261-53335261 on Build GRCh38
rs1048027619 Chr.6: 53335172-53335172 on Build GRCh38
rs1005042320 Chr.6: 53335122-53335122 on Build GRCh38
rs566639372 Chr.6: 53335569-53335569 on Build GRCh38
rs920283082 Chr.6: 53335135-53335135 on Build GRCh38
rs548394243 Chr.6: 53335514-53335514 on Build GRCh38
rs550663098 Chr.6: 53335349-53335349 on Build GRCh38
rs77033722 Chr.6: 53335289-53335289 on Build GRCh38
rs943171736 Chr.6: 53335123-53335123 on Build GRCh38
rs1023488419 Chr.6: 53335144-53335144 on Build GRCh38
rs768265364 Chr.6: 53335278-53335278 on Build GRCh38
rs994625642 Chr.6: 53335369-53335369 on Build GRCh38

View Full Product Details