Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90123886-90124435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P4
Entrez Gene ID
442523
Gene Name
DPY19L2 pseudogene 4
Gene Aliases
-
UniGene
Hs.406964
Gene Chromosome Location
Chr.7: 90119400-90125600 on Build GRCh38
Transcripts
NR_003551.1 NR_110029.2

Gene Symbol
STEAP2-AS1
Entrez Gene ID
100874100
Gene Name
STEAP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.573339
Gene Chromosome Location
Chr.7: 89882353-90211635 on Build GRCh38
Transcripts
NR_003551.1 NR_110029.2

SNP Detail

SNP ID Chromosome Location
rs545191778 Chr.7: 90124279-90124279 on Build GRCh38
rs895744374 Chr.7: 90124190-90124190 on Build GRCh38
rs537173970 Chr.7: 90124042-90124042 on Build GRCh38
rs994585630 Chr.7: 90124304-90124304 on Build GRCh38
rs577879591 Chr.7: 90124253-90124253 on Build GRCh38
rs774065501 Chr.7: 90124002-90124004 on Build GRCh38
rs973456871 Chr.7: 90123982-90123982 on Build GRCh38
rs1042752498 Chr.7: 90124095-90124095 on Build GRCh38
rs1012718279 Chr.7: 90124205-90124205 on Build GRCh38
rs974887945 Chr.7: 90124072-90124072 on Build GRCh38
rs1025995426 Chr.7: 90123997-90123997 on Build GRCh38
rs146404803 Chr.7: 90123998-90123998 on Build GRCh38
rs187617894 Chr.7: 90124013-90124013 on Build GRCh38
rs1026504532 Chr.7: 90124342-90124342 on Build GRCh38
rs566365206 Chr.7: 90123914-90123914 on Build GRCh38
rs5007219 Chr.7: 90124155-90124155 on Build GRCh38
rs946839054 Chr.7: 90124091-90124091 on Build GRCh38
rs915285742 Chr.7: 90124077-90124077 on Build GRCh38
rs762861139 Chr.7: 90123985-90123989 on Build GRCh38
rs373128469 Chr.7: 90124025-90124025 on Build GRCh38
rs5007218 Chr.7: 90124156-90124156 on Build GRCh38
rs963163333 Chr.7: 90123943-90123943 on Build GRCh38
rs552947791 Chr.7: 90124251-90124251 on Build GRCh38
rs569783455 Chr.7: 90124038-90124038 on Build GRCh38
rs1057064683 Chr.7: 90124136-90124136 on Build GRCh38
rs902498693 Chr.7: 90124106-90124106 on Build GRCh38
rs567596422 Chr.7: 90124135-90124135 on Build GRCh38
rs73710130 Chr.7: 90124214-90124214 on Build GRCh38
rs1022824382 Chr.7: 90124207-90124207 on Build GRCh38
rs943531156 Chr.7: 90124034-90124034 on Build GRCh38
rs5007222 Chr.7: 90124094-90124094 on Build GRCh38
rs987720661 Chr.7: 90124011-90124011 on Build GRCh38
rs950408466 Chr.7: 90124362-90124362 on Build GRCh38

View Full Product Details