Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57516692-57516847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDIT3
Entrez Gene ID
1649
Gene Name
DNA damage inducible transcript 3
Gene Aliases
CEBPZ, CHOP, CHOP-10, CHOP10, GADD153
UniGene
Hs.505777
Gene Chromosome Location
Chr.12: 57516588-57520517 on Build GRCh38
Transcripts
NM_001195053.1 NM_001195054.1 NM_001195055.1 NM_001195056.1 NM_001195057.1 NM_004083.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6227645 c.548T>C p.I183T Chr.12: 57516840-57516840 on Build GRCh38
6227646 c.479T>C p.I160T Chr.12: 57516840-57516840 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1057229054 Chr.12: 57516812-57516812 on Build GRCh38
rs780733767 Chr.12: 57516767-57516767 on Build GRCh38
rs182097761 Chr.12: 57516716-57516716 on Build GRCh38
rs981054561 Chr.12: 57516817-57516817 on Build GRCh38
rs971306627 Chr.12: 57516805-57516805 on Build GRCh38
rs777269541 Chr.12: 57516797-57516804 on Build GRCh38
rs368301051 Chr.12: 57516786-57516786 on Build GRCh38
rs11547020 Chr.12: 57516765-57516765 on Build GRCh38
rs755753113 Chr.12: 57516783-57516783 on Build GRCh38
rs921111628 Chr.12: 57516734-57516735 on Build GRCh38
rs747778183 Chr.12: 57516779-57516779 on Build GRCh38
rs745766747 Chr.12: 57516804-57516804 on Build GRCh38
rs772089247 Chr.12: 57516807-57516807 on Build GRCh38
rs965875703 Chr.12: 57516723-57516723 on Build GRCh38
rs774493745 Chr.12: 57516798-57516798 on Build GRCh38
rs1027897283 Chr.12: 57516741-57516741 on Build GRCh38
rs1022807484 Chr.12: 57516800-57516800 on Build GRCh38
rs563855250 Chr.12: 57516739-57516739 on Build GRCh38
rs878950898 Chr.12: 57516757-57516757 on Build GRCh38
rs975219878 Chr.12: 57516732-57516733 on Build GRCh38

View Full Product Details