Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1438626-1438889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS15
Entrez Gene ID
6209
Gene Name
ribosomal protein S15
Gene Aliases
RIG, S15
UniGene
Hs.406683
Gene Chromosome Location
Chr.19: 1438364-1440497 on Build GRCh38
Transcripts
NM_001018.4 NM_001308226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370006 c.18G>T p.Q6H Chr.19: 1438821-1438821 on Build GRCh38
5655334 c.73C>T p.L25L Chr.19: 1438876-1438876 on Build GRCh38
5966264 c.45C>A p.F15L Chr.19: 1438848-1438848 on Build GRCh38
3756371 c.63C>T p.D21D Chr.19: 1438866-1438866 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1026595402 Chr.19: 1438693-1438693 on Build GRCh38
rs899174994 Chr.19: 1438679-1438679 on Build GRCh38
rs949885521 Chr.19: 1438751-1438751 on Build GRCh38
rs1028179492 Chr.19: 1438721-1438721 on Build GRCh38
rs769722014 Chr.19: 1438869-1438869 on Build GRCh38
rs988733502 Chr.19: 1438748-1438748 on Build GRCh38
rs780984042 Chr.19: 1438762-1438762 on Build GRCh38
rs978929237 Chr.19: 1438739-1438739 on Build GRCh38
rs895292670 Chr.19: 1438669-1438669 on Build GRCh38
rs754654464 Chr.19: 1438839-1438839 on Build GRCh38
rs1038214393 Chr.19: 1438809-1438809 on Build GRCh38
rs765902169 Chr.19: 1438740-1438740 on Build GRCh38
rs749964511 Chr.19: 1438684-1438684 on Build GRCh38
rs1002703090 Chr.19: 1438647-1438647 on Build GRCh38
rs934347750 Chr.19: 1438742-1438742 on Build GRCh38
rs772956934 Chr.19: 1438820-1438822 on Build GRCh38
rs762558851 Chr.19: 1438797-1438797 on Build GRCh38
rs1016233557 Chr.19: 1438653-1438653 on Build GRCh38
rs949512336 Chr.19: 1438678-1438678 on Build GRCh38
rs977783932 Chr.19: 1438646-1438646 on Build GRCh38
rs764754977 Chr.19: 1438828-1438828 on Build GRCh38
rs7251165 Chr.19: 1438866-1438866 on Build GRCh38
rs769723903 Chr.19: 1438765-1438765 on Build GRCh38
rs938080461 Chr.19: 1438801-1438801 on Build GRCh38
rs1046944797 Chr.19: 1438691-1438691 on Build GRCh38
rs780893945 Chr.19: 1438860-1438860 on Build GRCh38
rs148070085 Chr.19: 1438833-1438833 on Build GRCh38
rs753116376 Chr.19: 1438821-1438821 on Build GRCh38
rs757963954 Chr.19: 1438834-1438834 on Build GRCh38
rs779549419 Chr.19: 1438835-1438835 on Build GRCh38
rs532256746 Chr.19: 1438814-1438814 on Build GRCh38
rs964457594 Chr.19: 1438718-1438718 on Build GRCh38
rs751171632 Chr.19: 1438836-1438836 on Build GRCh38
rs778500677 Chr.19: 1438648-1438648 on Build GRCh38
rs995361749 Chr.19: 1438720-1438720 on Build GRCh38
rs377627432 Chr.19: 1438786-1438786 on Build GRCh38
rs540842874 Chr.19: 1438715-1438715 on Build GRCh38
rs1044170343 Chr.19: 1438779-1438779 on Build GRCh38
rs564942317 Chr.19: 1438749-1438749 on Build GRCh38
rs573458233 Chr.19: 1438692-1438692 on Build GRCh38
rs1055313001 Chr.19: 1438667-1438667 on Build GRCh38
rs555270729 Chr.19: 1438673-1438673 on Build GRCh38
rs771030446 Chr.19: 1438787-1438787 on Build GRCh38
rs534942408 Chr.19: 1438695-1438695 on Build GRCh38
rs374430711 Chr.19: 1438661-1438661 on Build GRCh38
rs761207093 Chr.19: 1438827-1438827 on Build GRCh38
rs759700768 Chr.19: 1438795-1438795 on Build GRCh38
rs746748321 Chr.19: 1438697-1438697 on Build GRCh38
rs774686705 Chr.19: 1438791-1438791 on Build GRCh38
rs1019822754 Chr.19: 1438757-1438757 on Build GRCh38
rs768544407 Chr.19: 1438709-1438709 on Build GRCh38

View Full Product Details