Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100991047-100991280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC12
Entrez Gene ID
10071
Gene Name
mucin 12, cell surface associated
Gene Aliases
MUC-11, MUC-12, MUC11
UniGene
Hs.489355
Gene Chromosome Location
Chr.7: 100969623-101018949 on Build GRCh38
Transcripts
NM_001164462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6190686 c.661T>A p.S221T Chr.7: 100991224-100991224 on Build GRCh38
5224369 c.1073C>T p.S358L Chr.7: 100991207-100991207 on Build GRCh38
4390846 c.495A>C p.S165S Chr.7: 100991058-100991058 on Build GRCh38
3081462 c.533T>C p.I178T Chr.7: 100991096-100991096 on Build GRCh38
1083216 c.703C>T p.R235C Chr.7: 100991266-100991266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371891242 Chr.7: 100991159-100991159 on Build GRCh38
rs907690064 Chr.7: 100991110-100991110 on Build GRCh38
rs1021790444 Chr.7: 100991213-100991213 on Build GRCh38
rs769176549 Chr.7: 100991087-100991087 on Build GRCh38
rs767638861 Chr.7: 100991257-100991257 on Build GRCh38
rs904193031 Chr.7: 100991240-100991240 on Build GRCh38
rs764486495 Chr.7: 100991108-100991108 on Build GRCh38
rs879151885 Chr.7: 100991175-100991175 on Build GRCh38
rs372961064 Chr.7: 100991163-100991163 on Build GRCh38
rs973219793 Chr.7: 100991118-100991118 on Build GRCh38
rs532704724 Chr.7: 100991245-100991245 on Build GRCh38
rs577415446 Chr.7: 100991127-100991127 on Build GRCh38
rs759776287 Chr.7: 100991252-100991252 on Build GRCh38
rs771764880 Chr.7: 100991143-100991143 on Build GRCh38
rs770482039 Chr.7: 100991169-100991169 on Build GRCh38
rs183463642 Chr.7: 100991150-100991150 on Build GRCh38
rs565524249 Chr.7: 100991203-100991203 on Build GRCh38
rs774018507 Chr.7: 100991100-100991100 on Build GRCh38
rs61746950 Chr.7: 100991128-100991128 on Build GRCh38
rs567173084 Chr.7: 100991088-100991088 on Build GRCh38
rs763779692 Chr.7: 100991096-100991096 on Build GRCh38
rs773904899 Chr.7: 100991216-100991216 on Build GRCh38
rs117301718 Chr.7: 100991105-100991105 on Build GRCh38
rs767869817 Chr.7: 100991082-100991082 on Build GRCh38
rs940149062 Chr.7: 100991117-100991117 on Build GRCh38
rs919975714 Chr.7: 100991123-100991123 on Build GRCh38
rs532066040 Chr.7: 100991165-100991165 on Build GRCh38
rs992637138 Chr.7: 100991106-100991106 on Build GRCh38
rs542581157 Chr.7: 100991161-100991161 on Build GRCh38
rs61746936 Chr.7: 100991075-100991075 on Build GRCh38
rs745589585 Chr.7: 100991187-100991187 on Build GRCh38
rs746487768 Chr.7: 100991074-100991074 on Build GRCh38
rs1034230127 Chr.7: 100991099-100991099 on Build GRCh38
rs61746937 Chr.7: 100991077-100991077 on Build GRCh38
rs374128187 Chr.7: 100991093-100991093 on Build GRCh38
rs556543189 Chr.7: 100991164-100991164 on Build GRCh38
rs199521685 Chr.7: 100991122-100991122 on Build GRCh38
rs891947782 Chr.7: 100991209-100991209 on Build GRCh38
rs766686439 Chr.7: 100991152-100991152 on Build GRCh38
rs778509336 Chr.7: 100991173-100991173 on Build GRCh38
rs370156782 Chr.7: 100991242-100991242 on Build GRCh38
rs6980143 Chr.7: 100991195-100991195 on Build GRCh38
rs770285254 Chr.7: 100991211-100991211 on Build GRCh38
rs370245340 Chr.7: 100991111-100991111 on Build GRCh38
rs763093474 Chr.7: 100991224-100991224 on Build GRCh38
rs776987015 Chr.7: 100991109-100991109 on Build GRCh38
rs1001167901 Chr.7: 100991244-100991244 on Build GRCh38
rs747393179 Chr.7: 100991167-100991167 on Build GRCh38
rs760282588 Chr.7: 100991083-100991083 on Build GRCh38

View Full Product Details