Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117186485-117186758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIDT2
Entrez Gene ID
51092
Gene Name
SID1 transmembrane family member 2
Gene Aliases
CGI-40
UniGene
Hs.712144
Gene Chromosome Location
Chr.11: 117179223-117197445 on Build GRCh38
Transcripts
NM_001040455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923403 c.998G>A p.R333Q Chr.11: 117186619-117186619 on Build GRCh38
6229686 c.1015G>A p.G339S Chr.11: 117186636-117186636 on Build GRCh38
2104453 c.977C>G p.T326S Chr.11: 117186598-117186598 on Build GRCh38
5780238 c.998G>A p.R333Q Chr.11: 117186619-117186619 on Build GRCh38
4184670 c.977C>G p.T326S Chr.11: 117186598-117186598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146800220 Chr.11: 117186619-117186619 on Build GRCh38
rs757921535 Chr.11: 117186618-117186618 on Build GRCh38
rs554490664 Chr.11: 117186679-117186679 on Build GRCh38
rs745911846 Chr.11: 117186685-117186686 on Build GRCh38
rs1014030086 Chr.11: 117186535-117186535 on Build GRCh38
rs896625762 Chr.11: 117186523-117186523 on Build GRCh38
rs569978978 Chr.11: 117186611-117186611 on Build GRCh38
rs139398188 Chr.11: 117186625-117186625 on Build GRCh38
rs568971451 Chr.11: 117186586-117186588 on Build GRCh38
rs371178656 Chr.11: 117186663-117186663 on Build GRCh38
rs774203830 Chr.11: 117186686-117186686 on Build GRCh38
rs372933306 Chr.11: 117186580-117186580 on Build GRCh38
rs765106930 Chr.11: 117186687-117186687 on Build GRCh38
rs771219796 Chr.11: 117186595-117186595 on Build GRCh38
rs538772152 Chr.11: 117186602-117186602 on Build GRCh38
rs763769550 Chr.11: 117186605-117186605 on Build GRCh38
rs748579710 Chr.11: 117186542-117186542 on Build GRCh38
rs972198716 Chr.11: 117186666-117186666 on Build GRCh38
rs367732298 Chr.11: 117186661-117186661 on Build GRCh38
rs753718397 Chr.11: 117186608-117186608 on Build GRCh38
rs201083017 Chr.11: 117186620-117186620 on Build GRCh38
rs774738797 Chr.11: 117186599-117186599 on Build GRCh38
rs143587311 Chr.11: 117186613-117186613 on Build GRCh38
rs749334808 Chr.11: 117186647-117186647 on Build GRCh38
rs199968814 Chr.11: 117186662-117186662 on Build GRCh38
rs770391096 Chr.11: 117186554-117186554 on Build GRCh38
rs536688630 Chr.11: 117186520-117186520 on Build GRCh38
rs375393077 Chr.11: 117186581-117186581 on Build GRCh38
rs781394867 Chr.11: 117186633-117186633 on Build GRCh38
rs550199653 Chr.11: 117186529-117186529 on Build GRCh38
rs756065989 Chr.11: 117186641-117186641 on Build GRCh38
rs745794959 Chr.11: 117186635-117186635 on Build GRCh38
rs1022680216 Chr.11: 117186562-117186562 on Build GRCh38
rs761678268 Chr.11: 117186686-117186686 on Build GRCh38
rs143730905 Chr.11: 117186578-117186578 on Build GRCh38
rs763130161 Chr.11: 117186587-117186587 on Build GRCh38
rs776288968 Chr.11: 117186681-117186681 on Build GRCh38

View Full Product Details