Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121965319-121965550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSMCE4A
Entrez Gene ID
54780
Gene Name
NSE4 homolog A, SMC5-SMC6 complex component
Gene Aliases
C10orf86, NS4EA, NSE4A
UniGene
Hs.258798
Gene Chromosome Location
Chr.10: 121957088-121975228 on Build GRCh38
Transcripts
NM_001167865.1 NM_017615.2 XM_005269929.4 XM_005269930.3 XM_011539910.2 XM_017016359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6455906 c.703C>T p.R235C Chr.10: 121965336-121965336 on Build GRCh38
3686544 c.694C>T p.R232* Chr.10: 121965345-121965345 on Build GRCh38
5790951 c.673G>T p.E225* Chr.10: 121965366-121965366 on Build GRCh38
5826634 c.694C>T p.R232* Chr.10: 121965345-121965345 on Build GRCh38
2056420 c.703C>T p.R235C Chr.10: 121965336-121965336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753033931 Chr.10: 121965410-121965411 on Build GRCh38
rs766238310 Chr.10: 121965411-121965411 on Build GRCh38
rs753537095 Chr.10: 121965431-121965431 on Build GRCh38
rs368853068 Chr.10: 121965370-121965370 on Build GRCh38
rs750099730 Chr.10: 121965387-121965396 on Build GRCh38
rs148221460 Chr.10: 121965375-121965375 on Build GRCh38
rs949336217 Chr.10: 121965469-121965469 on Build GRCh38
rs746126284 Chr.10: 121965357-121965357 on Build GRCh38
rs1006767945 Chr.10: 121965366-121965366 on Build GRCh38
rs149089631 Chr.10: 121965348-121965348 on Build GRCh38
rs949647610 Chr.10: 121965472-121965472 on Build GRCh38
rs371992226 Chr.10: 121965376-121965376 on Build GRCh38
rs543882795 Chr.10: 121965374-121965374 on Build GRCh38
rs760227908 Chr.10: 121965410-121965410 on Build GRCh38
rs771346068 Chr.10: 121965344-121965344 on Build GRCh38
rs76497370 Chr.10: 121965411-121965411 on Build GRCh38
rs902480973 Chr.10: 121965514-121965514 on Build GRCh38
rs778278410 Chr.10: 121965433-121965436 on Build GRCh38
rs12259144 Chr.10: 121965433-121965433 on Build GRCh38
rs1046646945 Chr.10: 121965492-121965492 on Build GRCh38
rs765452015 Chr.10: 121965459-121965459 on Build GRCh38
rs925035430 Chr.10: 121965377-121965377 on Build GRCh38
rs935355628 Chr.10: 121965516-121965516 on Build GRCh38
rs769856609 Chr.10: 121965359-121965359 on Build GRCh38
rs942182950 Chr.10: 121965465-121965465 on Build GRCh38
rs143064887 Chr.10: 121965369-121965369 on Build GRCh38
rs990975488 Chr.10: 121965468-121965468 on Build GRCh38
rs1047621771 Chr.10: 121965496-121965496 on Build GRCh38
rs758957024 Chr.10: 121965430-121965433 on Build GRCh38
rs1032437938 Chr.10: 121965345-121965345 on Build GRCh38

View Full Product Details