Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137254784-137255045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTN
Entrez Gene ID
5764
Gene Name
pleiotrophin
Gene Aliases
HARP, HB-GAM, HBBM, HBGF-8, HBGF8, HBNF, HBNF-1, NEGF1, OSF-1
UniGene
Hs.371249
Gene Chromosome Location
Chr.7: 137227341-137343865 on Build GRCh38
Transcripts
NM_001321386.1 NM_001321387.1 NM_002825.5 XM_017012473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6758752 c.55G>A p.A19T Chr.7: 137254919-137254919 on Build GRCh38
6758751 c.28C>T p.R10C Chr.7: 137254946-137254946 on Build GRCh38
3878404 c.48C>A p.A16A Chr.7: 137254926-137254926 on Build GRCh38
1085939 c.94G>T p.A32S Chr.7: 137254880-137254880 on Build GRCh38
5062212 c.109A>T p.K37* Chr.7: 137254865-137254865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375813942 Chr.7: 137254925-137254925 on Build GRCh38
rs73162498 Chr.7: 137254830-137254830 on Build GRCh38
rs772065463 Chr.7: 137254960-137254960 on Build GRCh38
rs761781300 Chr.7: 137254959-137254959 on Build GRCh38
rs571710032 Chr.7: 137255006-137255006 on Build GRCh38
rs777658614 Chr.7: 137254907-137254907 on Build GRCh38
rs748720545 Chr.7: 137254875-137254875 on Build GRCh38
rs142905210 Chr.7: 137254942-137254942 on Build GRCh38
rs760269986 Chr.7: 137255003-137255003 on Build GRCh38
rs761399836 Chr.7: 137254819-137254819 on Build GRCh38
rs768633698 Chr.7: 137254945-137254945 on Build GRCh38
rs747136336 Chr.7: 137254919-137254919 on Build GRCh38
rs992636462 Chr.7: 137254996-137254996 on Build GRCh38
rs750484865 Chr.7: 137254993-137254993 on Build GRCh38
rs377653677 Chr.7: 137254828-137254828 on Build GRCh38
rs774483255 Chr.7: 137254816-137254816 on Build GRCh38
rs765924126 Chr.7: 137255004-137255004 on Build GRCh38
rs755670057 Chr.7: 137254831-137254831 on Build GRCh38
rs755383434 Chr.7: 137254850-137254850 on Build GRCh38
rs747162194 Chr.7: 137254979-137254987 on Build GRCh38
rs1006096197 Chr.7: 137254926-137254926 on Build GRCh38
rs776955119 Chr.7: 137254935-137254935 on Build GRCh38
rs537965312 Chr.7: 137254862-137254862 on Build GRCh38
rs758912047 Chr.7: 137254898-137254898 on Build GRCh38
rs759889347 Chr.7: 137255007-137255007 on Build GRCh38
rs754119600 Chr.7: 137254839-137254839 on Build GRCh38
rs1162122 Chr.7: 137254835-137254835 on Build GRCh38
rs529925609 Chr.7: 137254824-137254824 on Build GRCh38
rs200423495 Chr.7: 137254946-137254946 on Build GRCh38
rs1025087512 Chr.7: 137255014-137255014 on Build GRCh38
rs772554423 Chr.7: 137254978-137254978 on Build GRCh38
rs145058396 Chr.7: 137254815-137254815 on Build GRCh38

View Full Product Details