Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52757664-52757889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ONECUT1
Entrez Gene ID
3175
Gene Name
one cut homeobox 1
Gene Aliases
HNF-6, HNF6, HNF6A
UniGene
Hs.658573
Gene Chromosome Location
Chr.15: 52756963-52790264 on Build GRCh38
Transcripts
NM_004498.2 NR_073510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5604316 c.1151C>A p.T384K Chr.15: 52757802-52757802 on Build GRCh38
2217573 c.1225C>T p.R409C Chr.15: 52757728-52757728 on Build GRCh38
1708187 c.1216G>A p.E406K Chr.15: 52757737-52757737 on Build GRCh38
4055600 c.1226G>A p.R409H Chr.15: 52757727-52757727 on Build GRCh38
3502201 c.1193G>A p.R398Q Chr.15: 52757760-52757760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs60211775 Chr.15: 52757739-52757739 on Build GRCh38
rs746910544 Chr.15: 52757760-52757760 on Build GRCh38
rs767975108 Chr.15: 52757727-52757727 on Build GRCh38
rs749522422 Chr.15: 52757684-52757684 on Build GRCh38
rs201797520 Chr.15: 52757851-52757851 on Build GRCh38
rs200225368 Chr.15: 52757753-52757753 on Build GRCh38
rs761223829 Chr.15: 52757729-52757729 on Build GRCh38
rs867823975 Chr.15: 52757815-52757815 on Build GRCh38
rs200829865 Chr.15: 52757763-52757763 on Build GRCh38
rs781028194 Chr.15: 52757785-52757785 on Build GRCh38
rs749905495 Chr.15: 52757751-52757751 on Build GRCh38
rs764415210 Chr.15: 52757737-52757737 on Build GRCh38
rs776002886 Chr.15: 52757728-52757728 on Build GRCh38
rs756805415 Chr.15: 52757833-52757833 on Build GRCh38
rs896504428 Chr.15: 52757800-52757800 on Build GRCh38
rs185768697 Chr.15: 52757836-52757836 on Build GRCh38
rs936988820 Chr.15: 52757830-52757830 on Build GRCh38
rs764798477 Chr.15: 52757750-52757750 on Build GRCh38
rs60181666 Chr.15: 52757803-52757803 on Build GRCh38
rs144952226 Chr.15: 52757786-52757786 on Build GRCh38
rs759541215 Chr.15: 52757723-52757723 on Build GRCh38
rs951270492 Chr.15: 52757757-52757757 on Build GRCh38
rs754209416 Chr.15: 52757748-52757748 on Build GRCh38
rs181518771 Chr.15: 52757703-52757703 on Build GRCh38
rs1056451758 Chr.15: 52757795-52757795 on Build GRCh38
rs774432033 Chr.15: 52757718-52757718 on Build GRCh38
rs771119211 Chr.15: 52757819-52757819 on Build GRCh38
rs1046931876 Chr.15: 52757713-52757713 on Build GRCh38
rs776607333 Chr.15: 52757683-52757683 on Build GRCh38

View Full Product Details