Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48628031-48628304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPHK2
Entrez Gene ID
56848
Gene Name
sphingosine kinase 2
Gene Aliases
SK 2, SK-2, SPK 2, SPK-2
UniGene
Hs.528006
Gene Chromosome Location
Chr.19: 48619291-48630407 on Build GRCh38
Transcripts
NM_001204158.2 NM_001204159.2 NM_001204160.2 NM_001243876.1 NM_020126.4 XM_006723292.1 XM_011527133.1 XM_011527134.1 XM_017027008.1 XM_017027009.1 XM_017027010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6454049 c.634G>A p.G212R Chr.19: 48628055-48628055 on Build GRCh38
3764758 c.942+10G>T p.? Chr.19: 48628079-48628079 on Build GRCh38
5856327 c.1051C>T p.H351Y Chr.19: 48628270-48628270 on Build GRCh38
3726133 c.872+10G>A p.? Chr.19: 48628287-48628287 on Build GRCh38
3536591 c.715C>T p.P239S Chr.19: 48628228-48628228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757657024 Chr.19: 48628200-48628200 on Build GRCh38
rs1021623877 Chr.19: 48628158-48628158 on Build GRCh38
rs765185855 Chr.19: 48628073-48628073 on Build GRCh38
rs376253681 Chr.19: 48628269-48628269 on Build GRCh38
rs768509866 Chr.19: 48628107-48628107 on Build GRCh38
rs1043398066 Chr.19: 48628238-48628238 on Build GRCh38
rs970530397 Chr.19: 48628096-48628096 on Build GRCh38
rs746600744 Chr.19: 48628048-48628048 on Build GRCh38
rs748435598 Chr.19: 48628272-48628273 on Build GRCh38
rs765660290 Chr.19: 48628276-48628276 on Build GRCh38
rs754224418 Chr.19: 48628116-48628116 on Build GRCh38
rs116785119 Chr.19: 48628060-48628060 on Build GRCh38
rs768279647 Chr.19: 48628155-48628155 on Build GRCh38
rs775661505 Chr.19: 48628184-48628184 on Build GRCh38
rs760824600 Chr.19: 48628260-48628260 on Build GRCh38
rs746304674 Chr.19: 48628143-48628143 on Build GRCh38
rs570647736 Chr.19: 48628156-48628156 on Build GRCh38
rs761792067 Chr.19: 48628064-48628064 on Build GRCh38
rs757933167 Chr.19: 48628146-48628146 on Build GRCh38
rs372795593 Chr.19: 48628104-48628104 on Build GRCh38
rs754312390 Chr.19: 48628194-48628194 on Build GRCh38
rs142182513 Chr.19: 48628272-48628272 on Build GRCh38
rs982322064 Chr.19: 48628279-48628279 on Build GRCh38
rs759607232 Chr.19: 48628247-48628247 on Build GRCh38
rs754689223 Chr.19: 48628233-48628233 on Build GRCh38
rs779668124 Chr.19: 48628221-48628221 on Build GRCh38
rs752966284 Chr.19: 48628101-48628101 on Build GRCh38
rs764574964 Chr.19: 48628108-48628108 on Build GRCh38
rs765455864 Chr.19: 48628212-48628212 on Build GRCh38
rs188257436 Chr.19: 48628150-48628150 on Build GRCh38
rs757905699 Chr.19: 48628120-48628120 on Build GRCh38
rs778526255 Chr.19: 48628124-48628125 on Build GRCh38
rs750734391 Chr.19: 48628280-48628280 on Build GRCh38
rs960409787 Chr.19: 48628229-48628229 on Build GRCh38
rs769675804 Chr.19: 48628164-48628164 on Build GRCh38
rs747929600 Chr.19: 48628157-48628157 on Build GRCh38
rs900261772 Chr.19: 48628055-48628055 on Build GRCh38
rs761027008 Chr.19: 48628185-48628185 on Build GRCh38
rs751042287 Chr.19: 48628227-48628227 on Build GRCh38
rs867878766 Chr.19: 48628253-48628253 on Build GRCh38
rs770750657 Chr.19: 48628254-48628254 on Build GRCh38
rs147603863 Chr.19: 48628246-48628246 on Build GRCh38
rs767436801 Chr.19: 48628100-48628100 on Build GRCh38
rs774359148 Chr.19: 48628094-48628094 on Build GRCh38
rs531500911 Chr.19: 48628218-48628218 on Build GRCh38
rs759645029 Chr.19: 48628099-48628099 on Build GRCh38
rs1023080394 Chr.19: 48628085-48628085 on Build GRCh38
rs768800840 Chr.19: 48628268-48628268 on Build GRCh38
rs921657758 Chr.19: 48628273-48628273 on Build GRCh38
rs532706673 Chr.19: 48628248-48628248 on Build GRCh38
rs764407642 Chr.19: 48628193-48628193 on Build GRCh38
rs762730992 Chr.19: 48628170-48628170 on Build GRCh38
rs750974721 Chr.19: 48628213-48628213 on Build GRCh38
rs779393636 Chr.19: 48628148-48628148 on Build GRCh38
rs747738457 Chr.19: 48628239-48628239 on Build GRCh38
rs375999606 Chr.19: 48628216-48628216 on Build GRCh38
rs767665089 Chr.19: 48628171-48628171 on Build GRCh38
rs773212395 Chr.19: 48628165-48628165 on Build GRCh38
rs779463260 Chr.19: 48628135-48628135 on Build GRCh38
rs768473899 Chr.19: 48628054-48628054 on Build GRCh38
rs550024745 Chr.19: 48628242-48628242 on Build GRCh38

View Full Product Details