Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103368024-103368273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF5
Entrez Gene ID
6877
Gene Name
TATA-box binding protein associated factor 5
Gene Aliases
TAF(II)100, TAF2D, TAFII-100, TAFII100
UniGene
Hs.96103
Gene Chromosome Location
Chr.10: 103367953-103389065 on Build GRCh38
Transcripts
NM_006951.4 NM_139052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7392961 c.89A>G p.D30G Chr.10: 103368078-103368078 on Build GRCh38
6422071 c.256C>A p.P86T Chr.10: 103368245-103368245 on Build GRCh38
7331215 c.232G>T p.A78S Chr.10: 103368221-103368221 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs930776991 Chr.10: 103368134-103368134 on Build GRCh38
rs1049262949 Chr.10: 103368149-103368149 on Build GRCh38
rs755773467 Chr.10: 103368252-103368252 on Build GRCh38
rs776748330 Chr.10: 103368202-103368202 on Build GRCh38
rs763910237 Chr.10: 103368112-103368120 on Build GRCh38
rs779058784 Chr.10: 103368062-103368062 on Build GRCh38
rs763456502 Chr.10: 103368220-103368220 on Build GRCh38
rs968302166 Chr.10: 103368060-103368060 on Build GRCh38
rs919691215 Chr.10: 103368091-103368091 on Build GRCh38
rs775919402 Chr.10: 103368213-103368213 on Build GRCh38
rs928310747 Chr.10: 103368045-103368045 on Build GRCh38
rs981479032 Chr.10: 103368044-103368044 on Build GRCh38
rs747214067 Chr.10: 103368145-103368145 on Build GRCh38
rs1049400023 Chr.10: 103368225-103368225 on Build GRCh38
rs764402804 Chr.10: 103368235-103368235 on Build GRCh38
rs773141927 Chr.10: 103368093-103368093 on Build GRCh38
rs748798733 Chr.10: 103368051-103368051 on Build GRCh38
rs771931173 Chr.10: 103368072-103368072 on Build GRCh38
rs937296212 Chr.10: 103368208-103368230 on Build GRCh38
rs771079772 Chr.10: 103368178-103368178 on Build GRCh38
rs976646998 Chr.10: 103368179-103368179 on Build GRCh38
rs944929955 Chr.10: 103368162-103368162 on Build GRCh38
rs765991440 Chr.10: 103368256-103368256 on Build GRCh38
rs369610870 Chr.10: 103368246-103368246 on Build GRCh38
rs978368757 Chr.10: 103368140-103368140 on Build GRCh38
rs924170555 Chr.10: 103368143-103368143 on Build GRCh38
rs747940460 Chr.10: 103368065-103368065 on Build GRCh38
rs189802876 Chr.10: 103368059-103368059 on Build GRCh38
rs765722591 Chr.10: 103368212-103368212 on Build GRCh38
rs759573716 Chr.10: 103368204-103368204 on Build GRCh38
rs866783432 Chr.10: 103368239-103368239 on Build GRCh38
rs866160012 Chr.10: 103368251-103368251 on Build GRCh38
rs751948176 Chr.10: 103368244-103368244 on Build GRCh38

View Full Product Details