Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3823862-3824087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5S1
Entrez Gene ID
55317
Gene Name
adaptor related protein complex 5 sigma 1 subunit
Gene Aliases
C20orf29
UniGene
Hs.104806
Gene Chromosome Location
Chr.20: 3820524-3825307 on Build GRCh38
Transcripts
NM_001204446.1 NM_001204447.1 NM_018347.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1193110 c.186G>T p.E62D Chr.20: 3823880-3823880 on Build GRCh38
4411314 c.220C>T p.R74W Chr.20: 3823914-3823914 on Build GRCh38
6601816 c.292T>G p.F98V Chr.20: 3823986-3823986 on Build GRCh38
577430 c.296G>T p.R99L Chr.20: 3823990-3823990 on Build GRCh38
6601814 c.203A>T p.Q68L Chr.20: 3823897-3823897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368207793 Chr.20: 3823924-3823924 on Build GRCh38
rs768409747 Chr.20: 3823937-3823937 on Build GRCh38
rs756409558 Chr.20: 3824046-3824046 on Build GRCh38
rs372930691 Chr.20: 3824005-3824005 on Build GRCh38
rs770259675 Chr.20: 3823932-3823932 on Build GRCh38
rs751669142 Chr.20: 3823984-3823984 on Build GRCh38
rs771052011 Chr.20: 3824020-3824020 on Build GRCh38
rs752893710 Chr.20: 3823964-3823964 on Build GRCh38
rs754537686 Chr.20: 3824053-3824053 on Build GRCh38
rs138665829 Chr.20: 3823992-3823992 on Build GRCh38
rs372314865 Chr.20: 3823899-3823899 on Build GRCh38
rs200140340 Chr.20: 3823998-3823998 on Build GRCh38
rs775017338 Chr.20: 3823935-3823935 on Build GRCh38
rs146153842 Chr.20: 3823987-3823987 on Build GRCh38
rs200277002 Chr.20: 3823884-3823884 on Build GRCh38
rs545414217 Chr.20: 3824051-3824051 on Build GRCh38
rs533773124 Chr.20: 3823914-3823914 on Build GRCh38
rs941895697 Chr.20: 3824031-3824031 on Build GRCh38
rs373768834 Chr.20: 3823889-3823889 on Build GRCh38
rs577794844 Chr.20: 3823988-3823988 on Build GRCh38
rs771723692 Chr.20: 3823952-3823952 on Build GRCh38
rs543869928 Chr.20: 3823938-3823938 on Build GRCh38
rs750751392 Chr.20: 3823990-3823990 on Build GRCh38
rs143505011 Chr.20: 3824023-3824023 on Build GRCh38
rs766389023 Chr.20: 3823961-3823961 on Build GRCh38
rs761671910 Chr.20: 3823943-3823943 on Build GRCh38
rs760544175 Chr.20: 3824018-3824018 on Build GRCh38
rs149938656 Chr.20: 3823923-3823923 on Build GRCh38
rs778336117 Chr.20: 3824057-3824057 on Build GRCh38
rs778657964 Chr.20: 3823997-3823997 on Build GRCh38
rs764044225 Chr.20: 3824027-3824027 on Build GRCh38
rs746338807 Chr.20: 3823929-3823929 on Build GRCh38
rs750319537 Chr.20: 3823905-3823905 on Build GRCh38
rs764393681 Chr.20: 3823978-3823978 on Build GRCh38
rs148813629 Chr.20: 3823999-3823999 on Build GRCh38
rs748808302 Chr.20: 3823994-3823994 on Build GRCh38
rs754787054 Chr.20: 3823930-3823931 on Build GRCh38
rs751834246 Chr.20: 3824028-3824028 on Build GRCh38
rs767501628 Chr.20: 3823893-3823893 on Build GRCh38
rs752801894 Chr.20: 3823917-3823917 on Build GRCh38
rs777555624 Chr.20: 3824067-3824067 on Build GRCh38
rs369900734 Chr.20: 3823989-3823989 on Build GRCh38
rs376444614 Chr.20: 3823977-3823977 on Build GRCh38
rs780542185 Chr.20: 3824050-3824050 on Build GRCh38
rs747663126 Chr.20: 3824064-3824064 on Build GRCh38
rs1021749258 Chr.20: 3823962-3823962 on Build GRCh38
rs367607495 Chr.20: 3823890-3823890 on Build GRCh38
rs763102207 Chr.20: 3824026-3824026 on Build GRCh38
rs138798757 Chr.20: 3823925-3823925 on Build GRCh38
rs767719093 Chr.20: 3824042-3824042 on Build GRCh38
rs748789852 Chr.20: 3823936-3823936 on Build GRCh38
rs776654347 Chr.20: 3824022-3824022 on Build GRCh38
rs771918520 Chr.20: 3824065-3824065 on Build GRCh38
rs375668374 Chr.20: 3823996-3823996 on Build GRCh38
rs556362780 Chr.20: 3823986-3823986 on Build GRCh38
rs755138972 Chr.20: 3823913-3823913 on Build GRCh38
rs142312165 Chr.20: 3823926-3823926 on Build GRCh38
rs760744709 Chr.20: 3823960-3823960 on Build GRCh38
rs184260219 Chr.20: 3823891-3823891 on Build GRCh38
rs376165428 Chr.20: 3824043-3824043 on Build GRCh38
rs1036186969 Chr.20: 3824037-3824037 on Build GRCh38
rs994202221 Chr.20: 3823915-3823915 on Build GRCh38
rs761977201 Chr.20: 3824030-3824030 on Build GRCh38
rs373790111 Chr.20: 3823991-3823991 on Build GRCh38
rs1039422979 Chr.20: 3823900-3823900 on Build GRCh38
rs371588969 Chr.20: 3823959-3823959 on Build GRCh38
rs775142141 Chr.20: 3823885-3823885 on Build GRCh38
rs894441909 Chr.20: 3823939-3823939 on Build GRCh38

View Full Product Details