Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109314583-109314856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SORT1
Entrez Gene ID
6272
Gene Name
sortilin 1
Gene Aliases
Gp95, LDLCQ6, NT3, NTR3
UniGene
Hs.485195
Gene Chromosome Location
Chr.1: 109309565-109397945 on Build GRCh38
Transcripts
NM_001205228.1 NM_002959.6 XM_005271100.2 XM_005271101.2 XM_005271102.1 XM_006710812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4729099 c.2351G>T p.G784V Chr.1: 109314678-109314678 on Build GRCh38
4403543 c.2312T>G p.V771G Chr.1: 109314717-109314717 on Build GRCh38
1227153 c.2267C>A p.S756Y Chr.1: 109314762-109314762 on Build GRCh38
7080399 c.2298G>C p.L766F Chr.1: 109314731-109314731 on Build GRCh38
4020347 c.2265T>G p.N755K Chr.1: 109314764-109314764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758134766 Chr.1: 109314735-109314735 on Build GRCh38
rs754013986 Chr.1: 109314722-109314722 on Build GRCh38
rs770388154 Chr.1: 109314762-109314762 on Build GRCh38
rs957627546 Chr.1: 109314754-109314754 on Build GRCh38
rs766600918 Chr.1: 109314719-109314719 on Build GRCh38
rs369352027 Chr.1: 109314628-109314628 on Build GRCh38
rs868273309 Chr.1: 109314645-109314645 on Build GRCh38
rs1033287486 Chr.1: 109314615-109314615 on Build GRCh38
rs912906396 Chr.1: 109314807-109314807 on Build GRCh38
rs764837206 Chr.1: 109314699-109314699 on Build GRCh38
rs537323771 Chr.1: 109314610-109314610 on Build GRCh38
rs986531379 Chr.1: 109314833-109314833 on Build GRCh38
rs773202784 Chr.1: 109314642-109314642 on Build GRCh38
rs373252991 Chr.1: 109314689-109314689 on Build GRCh38
rs774754113 Chr.1: 109314712-109314712 on Build GRCh38
rs867940197 Chr.1: 109314759-109314759 on Build GRCh38
rs776216947 Chr.1: 109314674-109314674 on Build GRCh38
rs930749212 Chr.1: 109314821-109314821 on Build GRCh38
rs376200762 Chr.1: 109314715-109314715 on Build GRCh38
rs367920646 Chr.1: 109314739-109314739 on Build GRCh38
rs761237614 Chr.1: 109314818-109314818 on Build GRCh38
rs769472520 Chr.1: 109314803-109314803 on Build GRCh38
rs369998819 Chr.1: 109314740-109314740 on Build GRCh38
rs373721142 Chr.1: 109314813-109314813 on Build GRCh38
rs779694806 Chr.1: 109314613-109314613 on Build GRCh38
rs146391412 Chr.1: 109314804-109314804 on Build GRCh38
rs770443196 Chr.1: 109314666-109314666 on Build GRCh38
rs767045435 Chr.1: 109314824-109314824 on Build GRCh38
rs762168195 Chr.1: 109314805-109314805 on Build GRCh38
rs966978096 Chr.1: 109314822-109314822 on Build GRCh38
rs780648686 Chr.1: 109314789-109314789 on Build GRCh38
rs746451403 Chr.1: 109314643-109314643 on Build GRCh38
rs747971943 Chr.1: 109314733-109314733 on Build GRCh38
rs989180136 Chr.1: 109314647-109314647 on Build GRCh38
rs773802660 Chr.1: 109314817-109314817 on Build GRCh38
rs755208822 Chr.1: 109314728-109314728 on Build GRCh38
rs913761433 Chr.1: 109314679-109314679 on Build GRCh38
rs772036482 Chr.1: 109314639-109314639 on Build GRCh38

View Full Product Details