Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1212268-1212497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A19
Entrez Gene ID
340024
Gene Name
solute carrier family 6 member 19
Gene Aliases
B0AT1, HND
UniGene
Hs.481478
Gene Chromosome Location
Chr.5: 1201595-1225117 on Build GRCh38
Transcripts
NM_001003841.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5918812 c.580C>T p.Q194* Chr.5: 1212401-1212401 on Build GRCh38
5024602 c.660G>A p.G220G Chr.5: 1212481-1212481 on Build GRCh38
6870679 c.614G>A p.W205* Chr.5: 1212435-1212435 on Build GRCh38
5802908 c.485A>G p.Y162C Chr.5: 1212306-1212306 on Build GRCh38
3714974 c.619G>A p.V207I Chr.5: 1212440-1212440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751451573 Chr.5: 1212301-1212301 on Build GRCh38
rs958805228 Chr.5: 1212393-1212393 on Build GRCh38
rs752611481 Chr.5: 1212409-1212409 on Build GRCh38
rs773280349 Chr.5: 1212390-1212390 on Build GRCh38
rs778676022 Chr.5: 1212378-1212378 on Build GRCh38
rs143274116 Chr.5: 1212462-1212462 on Build GRCh38
rs763090941 Chr.5: 1212354-1212354 on Build GRCh38
rs1025097531 Chr.5: 1212346-1212346 on Build GRCh38
rs772410636 Chr.5: 1212320-1212320 on Build GRCh38
rs138326359 Chr.5: 1212408-1212408 on Build GRCh38
rs368260483 Chr.5: 1212302-1212302 on Build GRCh38
rs781251649 Chr.5: 1212303-1212303 on Build GRCh38
rs753844063 Chr.5: 1212479-1212479 on Build GRCh38
rs199734007 Chr.5: 1212375-1212375 on Build GRCh38
rs778170943 Chr.5: 1212472-1212472 on Build GRCh38
rs201070872 Chr.5: 1212461-1212461 on Build GRCh38
rs1021640221 Chr.5: 1212415-1212415 on Build GRCh38
rs748579288 Chr.5: 1212439-1212439 on Build GRCh38
rs777075988 Chr.5: 1212383-1212383 on Build GRCh38
rs1025188287 Chr.5: 1212358-1212358 on Build GRCh38
rs121434346 Chr.5: 1212338-1212338 on Build GRCh38
rs761159018 Chr.5: 1212324-1212324 on Build GRCh38
rs375770616 Chr.5: 1212368-1212368 on Build GRCh38
rs147837686 Chr.5: 1212353-1212353 on Build GRCh38
rs747642051 Chr.5: 1212313-1212313 on Build GRCh38
rs376080842 Chr.5: 1212463-1212463 on Build GRCh38
rs775407906 Chr.5: 1212469-1212469 on Build GRCh38
rs1048868710 Chr.5: 1212445-1212445 on Build GRCh38
rs766694319 Chr.5: 1212356-1212356 on Build GRCh38
rs868381615 Chr.5: 1212396-1212396 on Build GRCh38
rs1004372728 Chr.5: 1212342-1212342 on Build GRCh38
rs774676628 Chr.5: 1212394-1212394 on Build GRCh38
rs764090881 Chr.5: 1212478-1212478 on Build GRCh38
rs777423162 Chr.5: 1212370-1212370 on Build GRCh38
rs747984234 Chr.5: 1212352-1212352 on Build GRCh38
rs550182623 Chr.5: 1212416-1212416 on Build GRCh38
rs751302903 Chr.5: 1212359-1212359 on Build GRCh38
rs140812221 Chr.5: 1212323-1212323 on Build GRCh38
rs752112692 Chr.5: 1212288-1212288 on Build GRCh38
rs115871703 Chr.5: 1212376-1212376 on Build GRCh38
rs760595534 Chr.5: 1212470-1212470 on Build GRCh38
rs372554453 Chr.5: 1212319-1212319 on Build GRCh38
rs113531820 Chr.5: 1212471-1212471 on Build GRCh38
rs912406302 Chr.5: 1212404-1212404 on Build GRCh38
rs770276029 Chr.5: 1212440-1212440 on Build GRCh38
rs767533794 Chr.5: 1212361-1212361 on Build GRCh38
rs748669031 Chr.5: 1212385-1212385 on Build GRCh38
rs140343487 Chr.5: 1212379-1212379 on Build GRCh38
rs772485568 Chr.5: 1212464-1212464 on Build GRCh38
rs765323895 Chr.5: 1212349-1212349 on Build GRCh38
rs141487939 Chr.5: 1212360-1212360 on Build GRCh38
rs375231132 Chr.5: 1212386-1212386 on Build GRCh38
rs778387537 Chr.5: 1212430-1212430 on Build GRCh38
rs769094393 Chr.5: 1212382-1212382 on Build GRCh38
rs201304215 Chr.5: 1212421-1212421 on Build GRCh38
rs990531960 Chr.5: 1212402-1212402 on Build GRCh38
rs756830423 Chr.5: 1212423-1212423 on Build GRCh38
rs779727632 Chr.5: 1212298-1212298 on Build GRCh38
rs769457402 Chr.5: 1212314-1212314 on Build GRCh38
rs377603594 Chr.5: 1212431-1212431 on Build GRCh38
rs752701535 Chr.5: 1212364-1212364 on Build GRCh38
rs559288285 Chr.5: 1212442-1212442 on Build GRCh38
rs199580276 Chr.5: 1212291-1212291 on Build GRCh38
rs769021114 Chr.5: 1212327-1212327 on Build GRCh38
rs764184677 Chr.5: 1212417-1212417 on Build GRCh38
rs766491340 Chr.5: 1212391-1212391 on Build GRCh38
rs201694226 Chr.5: 1212316-1212316 on Build GRCh38
rs200488896 Chr.5: 1212330-1212330 on Build GRCh38

View Full Product Details