Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123286584-123286857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY5
Entrez Gene ID
111
Gene Name
adenylate cyclase 5
Gene Aliases
AC5, FDFM
UniGene
Hs.593292
Gene Chromosome Location
Chr.3: 123282296-123448632 on Build GRCh38
Transcripts
NM_001199642.1 NM_183357.2 XM_005247077.3 XM_005247078.1 XM_006713483.1 XM_006713484.1 XM_011512359.2 XM_011512360.2 XM_011512361.1 XM_017005638.1 XM_017005639.1 XM_017005640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4659919 c.3606C>T p.T1202T Chr.3: 123286736-123286736 on Build GRCh38
6544748 c.3645C>A p.P1215P Chr.3: 123286697-123286697 on Build GRCh38
3586461 c.3637G>A p.G1213S Chr.3: 123286705-123286705 on Build GRCh38
5474540 c.3613G>A p.V1205M Chr.3: 123286729-123286729 on Build GRCh38
5497724 c.3543C>T p.I1181I Chr.3: 123286799-123286799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758210719 Chr.3: 123286792-123286792 on Build GRCh38
rs772583101 Chr.3: 123286666-123286666 on Build GRCh38
rs998825622 Chr.3: 123286768-123286768 on Build GRCh38
rs746530195 Chr.3: 123286679-123286679 on Build GRCh38
rs867359393 Chr.3: 123286714-123286714 on Build GRCh38
rs774848359 Chr.3: 123286705-123286705 on Build GRCh38
rs148824947 Chr.3: 123286767-123286767 on Build GRCh38
rs971554194 Chr.3: 123286643-123286643 on Build GRCh38
rs780896412 Chr.3: 123286821-123286821 on Build GRCh38
rs138624072 Chr.3: 123286730-123286730 on Build GRCh38
rs192847448 Chr.3: 123286628-123286628 on Build GRCh38
rs774523939 Chr.3: 123286831-123286831 on Build GRCh38
rs778567256 Chr.3: 123286784-123286784 on Build GRCh38
rs775516514 Chr.3: 123286721-123286721 on Build GRCh38
rs76023261 Chr.3: 123286629-123286629 on Build GRCh38
rs769550444 Chr.3: 123286823-123286823 on Build GRCh38
rs745482851 Chr.3: 123286822-123286822 on Build GRCh38
rs867392033 Chr.3: 123286726-123286726 on Build GRCh38
rs750439767 Chr.3: 123286720-123286720 on Build GRCh38
rs776449908 Chr.3: 123286696-123286696 on Build GRCh38
rs943468765 Chr.3: 123286834-123286834 on Build GRCh38
rs200974373 Chr.3: 123286651-123286651 on Build GRCh38
rs201070773 Chr.3: 123286754-123286754 on Build GRCh38
rs748031112 Chr.3: 123286785-123286785 on Build GRCh38
rs766539908 Chr.3: 123286736-123286736 on Build GRCh38
rs764570316 Chr.3: 123286793-123286793 on Build GRCh38
rs770583839 Chr.3: 123286818-123286818 on Build GRCh38
rs746474779 Chr.3: 123286799-123286799 on Build GRCh38
rs762247893 Chr.3: 123286835-123286835 on Build GRCh38
rs534019110 Chr.3: 123286718-123286718 on Build GRCh38
rs1031954956 Chr.3: 123286621-123286621 on Build GRCh38
rs772112446 Chr.3: 123286639-123286639 on Build GRCh38
rs540772645 Chr.3: 123286706-123286706 on Build GRCh38
rs377309404 Chr.3: 123286697-123286697 on Build GRCh38
rs199751744 Chr.3: 123286632-123286632 on Build GRCh38
rs770637165 Chr.3: 123286692-123286692 on Build GRCh38
rs764340902 Chr.3: 123286700-123286700 on Build GRCh38
rs150164904 Chr.3: 123286712-123286712 on Build GRCh38

View Full Product Details