Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1916555-1916822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALML6
Entrez Gene ID
163688
Gene Name
calmodulin like 6
Gene Aliases
CAGLP
UniGene
Hs.85902
Gene Chromosome Location
Chr.1: 1913751-1917294 on Build GRCh38
Transcripts
NM_138705.2 XM_005244729.3 XM_011540841.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359325 c.259G>A p.G87R Chr.1: 1916757-1916757 on Build GRCh38
4538845 c.260G>A p.G87E Chr.1: 1916758-1916758 on Build GRCh38
5786197 c.219G>C p.E73D Chr.1: 1916581-1916581 on Build GRCh38
6619122 c.286G>T p.A96S Chr.1: 1916784-1916784 on Build GRCh38
3803163 c.321C>G p.N107K Chr.1: 1916819-1916819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781045096 Chr.1: 1916658-1916658 on Build GRCh38
rs759795676 Chr.1: 1916743-1916743 on Build GRCh38
rs201925985 Chr.1: 1916656-1916656 on Build GRCh38
rs768053464 Chr.1: 1916635-1916635 on Build GRCh38
rs772710272 Chr.1: 1916730-1916730 on Build GRCh38
rs1048142154 Chr.1: 1916719-1916719 on Build GRCh38
rs777591197 Chr.1: 1916583-1916583 on Build GRCh38
rs534957043 Chr.1: 1916681-1916681 on Build GRCh38
rs755058878 Chr.1: 1916650-1916650 on Build GRCh38
rs778639993 Chr.1: 1916772-1916772 on Build GRCh38
rs200800474 Chr.1: 1916792-1916792 on Build GRCh38
rs766538533 Chr.1: 1916645-1916645 on Build GRCh38
rs761141748 Chr.1: 1916734-1916734 on Build GRCh38
rs28670453 Chr.1: 1916741-1916741 on Build GRCh38
rs200415259 Chr.1: 1916767-1916767 on Build GRCh38
rs747586485 Chr.1: 1916690-1916690 on Build GRCh38
rs762396016 Chr.1: 1916616-1916616 on Build GRCh38
rs953793471 Chr.1: 1916780-1916780 on Build GRCh38
rs779623102 Chr.1: 1916717-1916717 on Build GRCh38
rs766741105 Chr.1: 1916739-1916739 on Build GRCh38
rs748702050 Chr.1: 1916633-1916634 on Build GRCh38
rs1016515809 Chr.1: 1916731-1916731 on Build GRCh38
rs975440430 Chr.1: 1916603-1916603 on Build GRCh38
rs1002892798 Chr.1: 1916687-1916687 on Build GRCh38
rs771081881 Chr.1: 1916720-1916721 on Build GRCh38
rs74773083 Chr.1: 1916758-1916758 on Build GRCh38
rs556966372 Chr.1: 1916733-1916733 on Build GRCh38
rs758437010 Chr.1: 1916751-1916751 on Build GRCh38
rs769445344 Chr.1: 1916602-1916602 on Build GRCh38
rs35930313 Chr.1: 1916578-1916578 on Build GRCh38
rs780957254 Chr.1: 1916591-1916591 on Build GRCh38
rs755857395 Chr.1: 1916704-1916704 on Build GRCh38
rs765287544 Chr.1: 1916744-1916744 on Build GRCh38
rs754041454 Chr.1: 1916647-1916647 on Build GRCh38
rs549849140 Chr.1: 1916632-1916632 on Build GRCh38
rs746939714 Chr.1: 1916588-1916588 on Build GRCh38
rs748940761 Chr.1: 1916720-1916720 on Build GRCh38
rs745607903 Chr.1: 1916600-1916600 on Build GRCh38
rs777038741 Chr.1: 1916796-1916796 on Build GRCh38
rs28634421 Chr.1: 1916721-1916721 on Build GRCh38
rs200019110 Chr.1: 1916655-1916655 on Build GRCh38
rs367726631 Chr.1: 1916771-1916771 on Build GRCh38
rs35971167 Chr.1: 1916786-1916786 on Build GRCh38
rs758893678 Chr.1: 1916649-1916650 on Build GRCh38
rs775171208 Chr.1: 1916604-1916604 on Build GRCh38
rs773725031 Chr.1: 1916638-1916638 on Build GRCh38
rs568745793 Chr.1: 1916711-1916735 on Build GRCh38
rs373605319 Chr.1: 1916649-1916649 on Build GRCh38
rs28547095 Chr.1: 1916587-1916587 on Build GRCh38
rs28529346 Chr.1: 1916678-1916678 on Build GRCh38
rs539887260 Chr.1: 1916791-1916791 on Build GRCh38
rs761053917 Chr.1: 1916639-1916639 on Build GRCh38
rs28472657 Chr.1: 1916682-1916682 on Build GRCh38
rs370943172 Chr.1: 1916626-1916626 on Build GRCh38
rs2803296 Chr.1: 1916670-1916670 on Build GRCh38
rs776875261 Chr.1: 1916757-1916757 on Build GRCh38
rs955551945 Chr.1: 1916609-1916610 on Build GRCh38
rs767467758 Chr.1: 1916753-1916753 on Build GRCh38
rs747603210 Chr.1: 1916728-1916728 on Build GRCh38
rs770748193 Chr.1: 1916589-1916589 on Build GRCh38
rs771659057 Chr.1: 1916781-1916781 on Build GRCh38
rs755043614 Chr.1: 1916579-1916579 on Build GRCh38
rs79330154 Chr.1: 1916689-1916689 on Build GRCh38
rs924881731 Chr.1: 1916621-1916621 on Build GRCh38
rs779504826 Chr.1: 1916632-1916634 on Build GRCh38
rs778064441 Chr.1: 1916650-1916650 on Build GRCh38
rs752782636 Chr.1: 1916748-1916748 on Build GRCh38
rs773899751 Chr.1: 1916723-1916723 on Build GRCh38
rs569744216 Chr.1: 1916634-1916634 on Build GRCh38
rs747484520 Chr.1: 1916788-1916788 on Build GRCh38
rs745697944 Chr.1: 1916662-1916662 on Build GRCh38

View Full Product Details