Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26336832-26337089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM1L
Entrez Gene ID
55057
Gene Name
absent in melanoma 1-like
Gene Aliases
CRYBG2
UniGene
Hs.128738
Gene Chromosome Location
Chr.1: 26321859-26354130 on Build GRCh38
Transcripts
NM_001039775.3 XM_005245918.2 XM_011541672.1 XM_011541673.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5661366 c.1254C>T p.G418G Chr.1: 26336876-26336876 on Build GRCh38
3746914 c.746G>A p.S249N Chr.1: 26336871-26336871 on Build GRCh38
5661365 c.741C>T p.G247G Chr.1: 26336876-26336876 on Build GRCh38
6597241 c.1174C>T p.L392L Chr.1: 26336956-26336956 on Build GRCh38
1182319 c.1190A>G p.D397G Chr.1: 26336940-26336940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775888758 Chr.1: 26336885-26336885 on Build GRCh38
rs150549145 Chr.1: 26336925-26336925 on Build GRCh38
rs776816705 Chr.1: 26336985-26336985 on Build GRCh38
rs761095916 Chr.1: 26336887-26336887 on Build GRCh38
rs368771562 Chr.1: 26336927-26336927 on Build GRCh38
rs546331332 Chr.1: 26336878-26336878 on Build GRCh38
rs376864521 Chr.1: 26337001-26337001 on Build GRCh38
rs546879753 Chr.1: 26337037-26337037 on Build GRCh38
rs542246875 Chr.1: 26337021-26337021 on Build GRCh38
rs373879831 Chr.1: 26336884-26336884 on Build GRCh38
rs1048058124 Chr.1: 26337068-26337068 on Build GRCh38
rs148396600 Chr.1: 26336966-26336966 on Build GRCh38
rs10751735 Chr.1: 26336871-26336871 on Build GRCh38
rs765982456 Chr.1: 26336972-26336986 on Build GRCh38
rs372233616 Chr.1: 26336971-26336971 on Build GRCh38
rs754344077 Chr.1: 26336913-26336913 on Build GRCh38
rs764547732 Chr.1: 26336907-26336907 on Build GRCh38
rs765778546 Chr.1: 26337000-26337000 on Build GRCh38
rs972502027 Chr.1: 26337022-26337022 on Build GRCh38
rs969996236 Chr.1: 26336950-26336950 on Build GRCh38
rs765437053 Chr.1: 26336923-26336923 on Build GRCh38
rs764538620 Chr.1: 26336979-26336979 on Build GRCh38
rs201912800 Chr.1: 26337012-26337012 on Build GRCh38
rs762265278 Chr.1: 26336929-26336930 on Build GRCh38
rs777630652 Chr.1: 26337025-26337025 on Build GRCh38
rs745367293 Chr.1: 26336957-26336957 on Build GRCh38
rs1017775554 Chr.1: 26336865-26336865 on Build GRCh38
rs762420598 Chr.1: 26336873-26336873 on Build GRCh38
rs762384758 Chr.1: 26336994-26336994 on Build GRCh38
rs866203655 Chr.1: 26337005-26337005 on Build GRCh38
rs770790761 Chr.1: 26336942-26336942 on Build GRCh38
rs576454165 Chr.1: 26337003-26337003 on Build GRCh38
rs377497312 Chr.1: 26336941-26336941 on Build GRCh38
rs561917384 Chr.1: 26337030-26337030 on Build GRCh38
rs757652571 Chr.1: 26336918-26336918 on Build GRCh38
rs753058632 Chr.1: 26336867-26336867 on Build GRCh38
rs746110728 Chr.1: 26336859-26336859 on Build GRCh38
rs200932297 Chr.1: 26336929-26336929 on Build GRCh38
rs149736171 Chr.1: 26336926-26336926 on Build GRCh38
rs748980242 Chr.1: 26337028-26337028 on Build GRCh38
rs895248223 Chr.1: 26337026-26337026 on Build GRCh38
rs755919191 Chr.1: 26336931-26336931 on Build GRCh38
rs987280993 Chr.1: 26336891-26336891 on Build GRCh38
rs201524654 Chr.1: 26336866-26336866 on Build GRCh38
rs771776325 Chr.1: 26336959-26336959 on Build GRCh38
rs759193403 Chr.1: 26336882-26336882 on Build GRCh38
rs766760208 Chr.1: 26337011-26337011 on Build GRCh38
rs369663154 Chr.1: 26336943-26336943 on Build GRCh38
rs1008181930 Chr.1: 26336855-26336855 on Build GRCh38
rs773173798 Chr.1: 26336872-26336872 on Build GRCh38
rs777383347 Chr.1: 26336939-26336939 on Build GRCh38
rs747965327 Chr.1: 26336870-26336870 on Build GRCh38
rs373472274 Chr.1: 26337052-26337052 on Build GRCh38
rs370637960 Chr.1: 26336879-26336879 on Build GRCh38

View Full Product Details