Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 39570455-39570688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YAE1D1
Entrez Gene ID
57002
Gene Name
Yae1 domain containing 1
Gene Aliases
C7orf36, GK003
UniGene
Hs.72168
Gene Chromosome Location
Chr.7: 39566376-39612089 on Build GRCh38
Transcripts
NM_001282446.1 NM_020192.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403824 c.218T>A p.I73N Chr.7: 39570594-39570594 on Build GRCh38
6824283 c.233G>A p.R78Q Chr.7: 39570609-39570609 on Build GRCh38
6824284 c.233G>A p.R78Q Chr.7: 39570609-39570609 on Build GRCh38
6403823 c.218T>A p.I73N Chr.7: 39570594-39570594 on Build GRCh38
6177908 c.130G>A p.E44K Chr.7: 39570506-39570506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747662467 Chr.7: 39570551-39570551 on Build GRCh38
rs766403561 Chr.7: 39570609-39570609 on Build GRCh38
rs6947660 Chr.7: 39570578-39570578 on Build GRCh38
rs905582741 Chr.7: 39570542-39570542 on Build GRCh38
rs142155970 Chr.7: 39570589-39570589 on Build GRCh38
rs375018997 Chr.7: 39570502-39570502 on Build GRCh38
rs2971 Chr.7: 39570642-39570642 on Build GRCh38
rs754749686 Chr.7: 39570623-39570623 on Build GRCh38
rs781572344 Chr.7: 39570558-39570558 on Build GRCh38
rs145248453 Chr.7: 39570560-39570560 on Build GRCh38
rs753697248 Chr.7: 39570615-39570615 on Build GRCh38
rs17171594 Chr.7: 39570561-39570561 on Build GRCh38
rs761821697 Chr.7: 39570601-39570601 on Build GRCh38
rs766238324 Chr.7: 39570528-39570528 on Build GRCh38
rs754733351 Chr.7: 39570547-39570547 on Build GRCh38
rs369272531 Chr.7: 39570541-39570541 on Build GRCh38
rs757881848 Chr.7: 39570554-39570554 on Build GRCh38
rs764290287 Chr.7: 39570499-39570499 on Build GRCh38
rs773138356 Chr.7: 39570605-39570605 on Build GRCh38
rs537049816 Chr.7: 39570513-39570513 on Build GRCh38
rs370264664 Chr.7: 39570592-39570592 on Build GRCh38
rs752348110 Chr.7: 39570627-39570627 on Build GRCh38
rs369233861 Chr.7: 39570553-39570553 on Build GRCh38
rs765008616 Chr.7: 39570624-39570624 on Build GRCh38
rs778649924 Chr.7: 39570548-39570548 on Build GRCh38
rs770168015 Chr.7: 39570574-39570574 on Build GRCh38
rs767689486 Chr.7: 39570603-39570603 on Build GRCh38
rs749597934 Chr.7: 39570584-39570584 on Build GRCh38
rs767389921 Chr.7: 39570514-39570514 on Build GRCh38
rs962192529 Chr.7: 39570494-39570494 on Build GRCh38
rs750326373 Chr.7: 39570521-39570521 on Build GRCh38
rs777381768 Chr.7: 39570643-39570643 on Build GRCh38
rs755983356 Chr.7: 39570527-39570527 on Build GRCh38
rs146248465 Chr.7: 39570608-39570608 on Build GRCh38

View Full Product Details