Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99516223-99516466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
UniGene
Hs.709288
Gene Chromosome Location
Chr.14: 99511266-99604390 on Build GRCh38
Transcripts
NM_001144995.1 XM_011536705.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470321 c.1100G>A p.R367Q Chr.14: 99516258-99516258 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547149751 Chr.14: 99516326-99516326 on Build GRCh38
rs950385168 Chr.14: 99516242-99516242 on Build GRCh38
rs547380744 Chr.14: 99516424-99516424 on Build GRCh38
rs557747266 Chr.14: 99516258-99516258 on Build GRCh38
rs753971811 Chr.14: 99516403-99516403 on Build GRCh38
rs745732459 Chr.14: 99516441-99516441 on Build GRCh38
rs1000781591 Chr.14: 99516337-99516337 on Build GRCh38
rs964002405 Chr.14: 99516404-99516404 on Build GRCh38
rs921842892 Chr.14: 99516316-99516316 on Build GRCh38
rs564103226 Chr.14: 99516446-99516446 on Build GRCh38
rs903801110 Chr.14: 99516330-99516330 on Build GRCh38
rs917642119 Chr.14: 99516298-99516298 on Build GRCh38
rs199770498 Chr.14: 99516249-99516249 on Build GRCh38
rs564789922 Chr.14: 99516379-99516379 on Build GRCh38
rs895888879 Chr.14: 99516401-99516401 on Build GRCh38
rs1052903549 Chr.14: 99516338-99516338 on Build GRCh38
rs755881941 Chr.14: 99516340-99516340 on Build GRCh38
rs1030386540 Chr.14: 99516360-99516360 on Build GRCh38
rs983184896 Chr.14: 99516243-99516243 on Build GRCh38
rs1043647141 Chr.14: 99516344-99516344 on Build GRCh38
rs578038492 Chr.14: 99516259-99516259 on Build GRCh38
rs953440909 Chr.14: 99516373-99516373 on Build GRCh38
rs372256838 Chr.14: 99516304-99516304 on Build GRCh38
rs570144396 Chr.14: 99516378-99516378 on Build GRCh38
rs1014399210 Chr.14: 99516439-99516439 on Build GRCh38
rs997892611 Chr.14: 99516348-99516348 on Build GRCh38
rs924071590 Chr.14: 99516411-99516411 on Build GRCh38
rs528814485 Chr.14: 99516389-99516389 on Build GRCh38
rs543582550 Chr.14: 99516272-99516272 on Build GRCh38

View Full Product Details