Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40793096-40793369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A15
Entrez Gene ID
10166
Gene Name
solute carrier family 25 member 15
Gene Aliases
D13S327, HHH, ORC1, ORNT1
UniGene
Hs.646645
Gene Chromosome Location
Chr.13: 40789411-40812460 on Build GRCh38
Transcripts
NM_014252.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6780541 c.45G>A p.A15A Chr.13: 40793271-40793271 on Build GRCh38
3732634 c.47G>A p.G16E Chr.13: 40793273-40793273 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs202247806 Chr.13: 40793270-40793270 on Build GRCh38
rs368684475 Chr.13: 40793195-40793195 on Build GRCh38
rs754981827 Chr.13: 40793221-40793221 on Build GRCh38
rs753693450 Chr.13: 40793322-40793322 on Build GRCh38
rs766122874 Chr.13: 40793210-40793210 on Build GRCh38
rs138184184 Chr.13: 40793308-40793308 on Build GRCh38
rs370279404 Chr.13: 40793246-40793246 on Build GRCh38
rs755980686 Chr.13: 40793202-40793202 on Build GRCh38
rs781526529 Chr.13: 40793242-40793242 on Build GRCh38
rs945840491 Chr.13: 40793154-40793154 on Build GRCh38
rs536293334 Chr.13: 40793177-40793177 on Build GRCh38
rs913082716 Chr.13: 40793137-40793137 on Build GRCh38
rs112276566 Chr.13: 40793160-40793160 on Build GRCh38
rs770009034 Chr.13: 40793251-40793251 on Build GRCh38
rs765829415 Chr.13: 40793299-40793299 on Build GRCh38
rs993092829 Chr.13: 40793123-40793123 on Build GRCh38
rs767210508 Chr.13: 40793197-40793197 on Build GRCh38
rs959931234 Chr.13: 40793121-40793121 on Build GRCh38
rs552939219 Chr.13: 40793219-40793219 on Build GRCh38
rs757399947 Chr.13: 40793241-40793241 on Build GRCh38
rs774823424 Chr.13: 40793292-40793292 on Build GRCh38
rs371080447 Chr.13: 40793185-40793185 on Build GRCh38
rs200958757 Chr.13: 40793271-40793271 on Build GRCh38
rs573033866 Chr.13: 40793244-40793244 on Build GRCh38
rs552201985 Chr.13: 40793298-40793298 on Build GRCh38
rs751757710 Chr.13: 40793239-40793239 on Build GRCh38
rs375382325 Chr.13: 40793186-40793186 on Build GRCh38
rs763063929 Chr.13: 40793318-40793318 on Build GRCh38
rs548145452 Chr.13: 40793124-40793124 on Build GRCh38
rs778928196 Chr.13: 40793237-40793237 on Build GRCh38
rs760271186 Chr.13: 40793315-40793315 on Build GRCh38
rs575712830 Chr.13: 40793293-40793293 on Build GRCh38
rs749585613 Chr.13: 40793279-40793279 on Build GRCh38
rs778530206 Chr.13: 40793200-40793200 on Build GRCh38
rs202193033 Chr.13: 40793316-40793316 on Build GRCh38
rs200219313 Chr.13: 40793287-40793287 on Build GRCh38

View Full Product Details