Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32333330-32333603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf75
Entrez Gene ID
64149
Gene Name
chromosome 17 open reading frame 75
Gene Aliases
NJMU-R1, SRI2
UniGene
Hs.655257
Gene Chromosome Location
Chr.17: 32328441-32350047 on Build GRCh38
Transcripts
NM_022344.3 XM_005258022.3 XM_017024940.1 XM_017024941.1

SNP Detail

SNP ID Chromosome Location
rs762542544 Chr.17: 32333490-32333490 on Build GRCh38
rs757129391 Chr.17: 32333570-32333571 on Build GRCh38
rs377285577 Chr.17: 32333408-32333408 on Build GRCh38
rs753821882 Chr.17: 32333555-32333555 on Build GRCh38
rs760614168 Chr.17: 32333566-32333566 on Build GRCh38
rs750946730 Chr.17: 32333380-32333380 on Build GRCh38
rs781020787 Chr.17: 32333444-32333444 on Build GRCh38
rs1051168446 Chr.17: 32333360-32333360 on Build GRCh38
rs761020816 Chr.17: 32333523-32333523 on Build GRCh38
rs753957093 Chr.17: 32333421-32333421 on Build GRCh38
rs745786199 Chr.17: 32333445-32333445 on Build GRCh38
rs375913041 Chr.17: 32333440-32333440 on Build GRCh38
rs372968809 Chr.17: 32333370-32333370 on Build GRCh38
rs779237571 Chr.17: 32333568-32333571 on Build GRCh38
rs779137410 Chr.17: 32333435-32333435 on Build GRCh38
rs1010819129 Chr.17: 32333552-32333552 on Build GRCh38
rs368558604 Chr.17: 32333511-32333511 on Build GRCh38
rs370875085 Chr.17: 32333438-32333438 on Build GRCh38
rs774119199 Chr.17: 32333509-32333509 on Build GRCh38
rs774801739 Chr.17: 32333485-32333485 on Build GRCh38
rs755053651 Chr.17: 32333557-32333557 on Build GRCh38
rs532151548 Chr.17: 32333500-32333500 on Build GRCh38
rs543645372 Chr.17: 32333467-32333467 on Build GRCh38
rs753998906 Chr.17: 32333567-32333568 on Build GRCh38
rs750868562 Chr.17: 32333517-32333517 on Build GRCh38
rs767983788 Chr.17: 32333512-32333512 on Build GRCh38
rs779012360 Chr.17: 32333561-32333561 on Build GRCh38
rs755097651 Chr.17: 32333430-32333430 on Build GRCh38
rs1032977409 Chr.17: 32333450-32333450 on Build GRCh38
rs965745194 Chr.17: 32333465-32333465 on Build GRCh38
rs766923184 Chr.17: 32333529-32333529 on Build GRCh38

View Full Product Details