Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 80133016-80133271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADNP2
Entrez Gene ID
22850
Gene Name
ADNP homeobox 2
Gene Aliases
ZNF508
UniGene
Hs.131915
Gene Chromosome Location
Chr.18: 80109031-80140346 on Build GRCh38
Transcripts
NM_014913.3 XM_005266656.4 XM_011525883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611446 c.188G>C p.G63A Chr.18: 80133182-80133182 on Build GRCh38
1711535 c.163G>A p.D55N Chr.18: 80133157-80133157 on Build GRCh38
5854497 c.131G>A p.G44E Chr.18: 80133125-80133125 on Build GRCh38
474066 c.109-2A>G p.? Chr.18: 80133101-80133101 on Build GRCh38
5428521 c.162T>A p.G54G Chr.18: 80133156-80133156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1064058 Chr.18: 80133156-80133156 on Build GRCh38
rs912805082 Chr.18: 80133192-80133192 on Build GRCh38
rs762944585 Chr.18: 80133092-80133092 on Build GRCh38
rs867194748 Chr.18: 80133179-80133179 on Build GRCh38
rs375480549 Chr.18: 80133230-80133230 on Build GRCh38
rs868532834 Chr.18: 80133057-80133057 on Build GRCh38
rs550719298 Chr.18: 80133109-80133109 on Build GRCh38
rs746030721 Chr.18: 80133123-80133123 on Build GRCh38
rs987085399 Chr.18: 80133191-80133191 on Build GRCh38
rs771123070 Chr.18: 80133082-80133082 on Build GRCh38
rs772604864 Chr.18: 80133056-80133056 on Build GRCh38
rs549163468 Chr.18: 80133188-80133188 on Build GRCh38
rs764149906 Chr.18: 80133113-80133113 on Build GRCh38
rs751344109 Chr.18: 80133131-80133131 on Build GRCh38
rs138962680 Chr.18: 80133144-80133144 on Build GRCh38
rs759867308 Chr.18: 80133087-80133087 on Build GRCh38
rs111782975 Chr.18: 80133196-80133196 on Build GRCh38
rs771193997 Chr.18: 80133208-80133208 on Build GRCh38
rs757459523 Chr.18: 80133080-80133080 on Build GRCh38
rs751676000 Chr.18: 80133079-80133080 on Build GRCh38
rs776722433 Chr.18: 80133086-80133086 on Build GRCh38
rs373903669 Chr.18: 80133090-80133090 on Build GRCh38
rs892694899 Chr.18: 80133112-80133112 on Build GRCh38
rs777923510 Chr.18: 80133076-80133077 on Build GRCh38
rs747393040 Chr.18: 80133078-80133078 on Build GRCh38
rs545406792 Chr.18: 80133088-80133088 on Build GRCh38
rs781663741 Chr.18: 80133217-80133217 on Build GRCh38
rs199819195 Chr.18: 80133155-80133155 on Build GRCh38
rs757124495 Chr.18: 80133140-80133140 on Build GRCh38
rs766918795 Chr.18: 80133098-80133098 on Build GRCh38
rs773522460 Chr.18: 80133063-80133063 on Build GRCh38
rs746055325 Chr.18: 80133219-80133219 on Build GRCh38
rs371673844 Chr.18: 80133052-80133052 on Build GRCh38
rs778264048 Chr.18: 80133202-80133202 on Build GRCh38
rs899984768 Chr.18: 80133077-80133077 on Build GRCh38

View Full Product Details