Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90905632-90905881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2A2
Entrez Gene ID
4122
Gene Name
mannosidase alpha class 2A member 2
Gene Aliases
MANA2X
UniGene
Hs.116459
Gene Chromosome Location
Chr.15: 90902218-90922585 on Build GRCh38
Transcripts
NM_001320977.1 NM_006122.3 NR_135502.1 NR_135503.1 NR_135504.1 XM_005254910.3 XM_017022184.1 XM_017022185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
135683 c.464C>T p.S155F Chr.15: 90905652-90905652 on Build GRCh38
265574 c.462_464delCTC p.S155delS Chr.15: 90905650-90905652 on Build GRCh38
702107 c.472C>T p.P158S Chr.15: 90905660-90905660 on Build GRCh38
4128585 c.444G>C p.W148C Chr.15: 90905632-90905632 on Build GRCh38
3420719 c.457G>A p.D153N Chr.15: 90905645-90905645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778592318 Chr.15: 90905665-90905665 on Build GRCh38
rs548215259 Chr.15: 90905784-90905784 on Build GRCh38
rs762129173 Chr.15: 90905715-90905715 on Build GRCh38
rs748811916 Chr.15: 90905662-90905662 on Build GRCh38
rs756597830 Chr.15: 90905759-90905759 on Build GRCh38
rs780677608 Chr.15: 90905761-90905761 on Build GRCh38
rs747773001 Chr.15: 90905796-90905796 on Build GRCh38
rs776471778 Chr.15: 90905687-90905687 on Build GRCh38
rs2072078 Chr.15: 90905785-90905785 on Build GRCh38
rs760532967 Chr.15: 90905677-90905677 on Build GRCh38
rs764099360 Chr.15: 90905702-90905702 on Build GRCh38
rs562495176 Chr.15: 90905667-90905667 on Build GRCh38
rs998787919 Chr.15: 90905775-90905775 on Build GRCh38
rs200800950 Chr.15: 90905661-90905661 on Build GRCh38
rs376743463 Chr.15: 90905799-90905799 on Build GRCh38
rs776578192 Chr.15: 90905846-90905846 on Build GRCh38
rs889286322 Chr.15: 90905705-90905705 on Build GRCh38
rs1031645545 Chr.15: 90905781-90905781 on Build GRCh38
rs745806630 Chr.15: 90905849-90905849 on Build GRCh38
rs374688808 Chr.15: 90905666-90905666 on Build GRCh38
rs770739979 Chr.15: 90905833-90905833 on Build GRCh38
rs768401548 Chr.15: 90905763-90905763 on Build GRCh38
rs1049196253 Chr.15: 90905700-90905700 on Build GRCh38
rs767916507 Chr.15: 90905747-90905747 on Build GRCh38
rs768714208 Chr.15: 90905857-90905857 on Build GRCh38
rs777449566 Chr.15: 90905812-90905812 on Build GRCh38
rs763045576 Chr.15: 90905701-90905701 on Build GRCh38
rs763490096 Chr.15: 90905727-90905728 on Build GRCh38
rs746772386 Chr.15: 90905832-90905832 on Build GRCh38
rs751746486 Chr.15: 90905713-90905713 on Build GRCh38
rs781697605 Chr.15: 90905656-90905656 on Build GRCh38
rs754462357 Chr.15: 90905766-90905766 on Build GRCh38
rs189880811 Chr.15: 90905750-90905750 on Build GRCh38
rs778303720 Chr.15: 90905795-90905795 on Build GRCh38
rs139539053 Chr.15: 90905653-90905653 on Build GRCh38
rs990194803 Chr.15: 90905790-90905790 on Build GRCh38
rs369206537 Chr.15: 90905686-90905686 on Build GRCh38
rs750890155 Chr.15: 90905657-90905657 on Build GRCh38
rs373319048 Chr.15: 90905762-90905762 on Build GRCh38
rs772998181 Chr.15: 90905671-90905671 on Build GRCh38

View Full Product Details