Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29575663-29575936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIT1
Entrez Gene ID
28964
Gene Name
GIT ArfGAP 1
Gene Aliases
-
UniGene
Hs.514051
Gene Chromosome Location
Chr.17: 29573469-29589592 on Build GRCh38
Transcripts
NM_001085454.1 NM_014030.3 XM_011524684.1 XM_011524685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1610029 c.1741C>T p.P581S Chr.17: 29575850-29575850 on Build GRCh38
7256972 c.1809C>T p.A603A Chr.17: 29575674-29575674 on Build GRCh38
4569395 c.1699T>C p.F567L Chr.17: 29575865-29575865 on Build GRCh38
4999228 c.1810G>A p.D604N Chr.17: 29575673-29575673 on Build GRCh38
6043066 c.1712C>T p.A571V Chr.17: 29575879-29575879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764425684 Chr.17: 29575786-29575786 on Build GRCh38
rs866501754 Chr.17: 29575689-29575689 on Build GRCh38
rs373310684 Chr.17: 29575733-29575733 on Build GRCh38
rs779235567 Chr.17: 29575842-29575842 on Build GRCh38
rs769775991 Chr.17: 29575864-29575864 on Build GRCh38
rs368859007 Chr.17: 29575790-29575790 on Build GRCh38
rs772780703 Chr.17: 29575744-29575744 on Build GRCh38
rs759556920 Chr.17: 29575769-29575769 on Build GRCh38
rs753751031 Chr.17: 29575691-29575691 on Build GRCh38
rs910138643 Chr.17: 29575834-29575834 on Build GRCh38
rs760191583 Chr.17: 29575747-29575747 on Build GRCh38
rs752519780 Chr.17: 29575706-29575706 on Build GRCh38
rs771739398 Chr.17: 29575736-29575736 on Build GRCh38
rs745368172 Chr.17: 29575827-29575827 on Build GRCh38
rs780353306 Chr.17: 29575804-29575805 on Build GRCh38
rs762146014 Chr.17: 29575895-29575895 on Build GRCh38
rs757419252 Chr.17: 29575812-29575812 on Build GRCh38
rs367764732 Chr.17: 29575915-29575915 on Build GRCh38
rs200441109 Chr.17: 29575720-29575720 on Build GRCh38
rs750544170 Chr.17: 29575916-29575916 on Build GRCh38
rs754857303 Chr.17: 29575693-29575693 on Build GRCh38
rs749601989 Chr.17: 29575735-29575735 on Build GRCh38
rs752572681 Chr.17: 29575781-29575781 on Build GRCh38
rs73986959 Chr.17: 29575734-29575734 on Build GRCh38
rs758201784 Chr.17: 29575784-29575784 on Build GRCh38
rs372896559 Chr.17: 29575894-29575894 on Build GRCh38
rs756664190 Chr.17: 29575823-29575823 on Build GRCh38
rs772727654 Chr.17: 29575685-29575685 on Build GRCh38
rs747090312 Chr.17: 29575725-29575725 on Build GRCh38
rs758929748 Chr.17: 29575856-29575856 on Build GRCh38
rs369858870 Chr.17: 29575712-29575712 on Build GRCh38
rs775998184 Chr.17: 29575766-29575766 on Build GRCh38
rs765038035 Chr.17: 29575770-29575770 on Build GRCh38
rs765971490 Chr.17: 29575763-29575763 on Build GRCh38
rs373891852 Chr.17: 29575686-29575686 on Build GRCh38
rs981847487 Chr.17: 29575782-29575782 on Build GRCh38
rs151175398 Chr.17: 29575747-29575762 on Build GRCh38
rs750445419 Chr.17: 29575819-29575819 on Build GRCh38
rs775510133 Chr.17: 29575872-29575872 on Build GRCh38
rs926315206 Chr.17: 29575783-29575783 on Build GRCh38
rs762974646 Chr.17: 29575879-29575879 on Build GRCh38
rs34015437 Chr.17: 29575889-29575890 on Build GRCh38
rs755275423 Chr.17: 29575813-29575813 on Build GRCh38
rs551350378 Chr.17: 29575713-29575713 on Build GRCh38
rs377012557 Chr.17: 29575833-29575833 on Build GRCh38
rs149623045 Chr.17: 29575690-29575690 on Build GRCh38
rs764051634 Chr.17: 29575886-29575886 on Build GRCh38
rs369909633 Chr.17: 29575732-29575732 on Build GRCh38
rs763846891 Chr.17: 29575849-29575849 on Build GRCh38
rs945790310 Chr.17: 29575708-29575708 on Build GRCh38
rs777388210 Chr.17: 29575721-29575721 on Build GRCh38
rs780495303 Chr.17: 29575831-29575831 on Build GRCh38
rs1032682645 Chr.17: 29575730-29575730 on Build GRCh38
rs199820269 Chr.17: 29575876-29575876 on Build GRCh38
rs567885599 Chr.17: 29575848-29575848 on Build GRCh38
rs756078582 Chr.17: 29575918-29575918 on Build GRCh38
rs753698037 Chr.17: 29575754-29575754 on Build GRCh38
rs756924655 Chr.17: 29575749-29575750 on Build GRCh38
rs781123598 Chr.17: 29575731-29575731 on Build GRCh38
rs1016291291 Chr.17: 29575756-29575756 on Build GRCh38
rs79321602 Chr.17: 29575729-29575729 on Build GRCh38
rs776228670 Chr.17: 29575852-29575852 on Build GRCh38
rs765144857 Chr.17: 29575702-29575702 on Build GRCh38

View Full Product Details