Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55354307-55354548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX6B2
Entrez Gene ID
125965
Gene Name
cytochrome c oxidase subunit 6B2
Gene Aliases
COXVIB2, CT59
UniGene
Hs.550544
Gene Chromosome Location
Chr.19: 55349702-55354814 on Build GRCh38
Transcripts
NM_144613.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634354 c.100C>A p.Q34K Chr.19: 55354422-55354422 on Build GRCh38
4899593 c.33G>A p.K11K Chr.19: 55354489-55354489 on Build GRCh38
6634355 c.96C>G p.C32W Chr.19: 55354426-55354426 on Build GRCh38
6086002 c.24G>T p.E8D Chr.19: 55354498-55354498 on Build GRCh38
335080 c.25C>A p.P9T Chr.19: 55354497-55354497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747694115 Chr.19: 55354416-55354424 on Build GRCh38
rs375376285 Chr.19: 55354493-55354493 on Build GRCh38
rs898856001 Chr.19: 55354350-55354350 on Build GRCh38
rs528839611 Chr.19: 55354498-55354498 on Build GRCh38
rs746728782 Chr.19: 55354407-55354407 on Build GRCh38
rs762135503 Chr.19: 55354458-55354458 on Build GRCh38
rs749286114 Chr.19: 55354518-55354518 on Build GRCh38
rs144019446 Chr.19: 55354433-55354433 on Build GRCh38
rs773385193 Chr.19: 55354526-55354526 on Build GRCh38
rs374797839 Chr.19: 55354461-55354461 on Build GRCh38
rs200588806 Chr.19: 55354360-55354360 on Build GRCh38
rs889269167 Chr.19: 55354333-55354333 on Build GRCh38
rs369960718 Chr.19: 55354522-55354522 on Build GRCh38
rs922040569 Chr.19: 55354343-55354343 on Build GRCh38
rs745372914 Chr.19: 55354497-55354497 on Build GRCh38
rs573125366 Chr.19: 55354331-55354331 on Build GRCh38
rs773993549 Chr.19: 55354457-55354457 on Build GRCh38
rs769864857 Chr.19: 55354440-55354440 on Build GRCh38
rs771696298 Chr.19: 55354420-55354420 on Build GRCh38
rs768448280 Chr.19: 55354451-55354451 on Build GRCh38
rs371406810 Chr.19: 55354474-55354474 on Build GRCh38
rs199913551 Chr.19: 55354529-55354529 on Build GRCh38
rs1007737132 Chr.19: 55354338-55354338 on Build GRCh38
rs201970981 Chr.19: 55354399-55354399 on Build GRCh38
rs754296563 Chr.19: 55354472-55354472 on Build GRCh38
rs775499700 Chr.19: 55354441-55354441 on Build GRCh38
rs376805143 Chr.19: 55354396-55354396 on Build GRCh38
rs762870786 Chr.19: 55354447-55354447 on Build GRCh38
rs200982714 Chr.19: 55354523-55354523 on Build GRCh38
rs777316035 Chr.19: 55354405-55354405 on Build GRCh38
rs199827691 Chr.19: 55354471-55354471 on Build GRCh38
rs770464196 Chr.19: 55354409-55354409 on Build GRCh38
rs138591719 Chr.19: 55354354-55354354 on Build GRCh38
rs772895257 Chr.19: 55354468-55354468 on Build GRCh38
rs1016008904 Chr.19: 55354342-55354342 on Build GRCh38
rs760682434 Chr.19: 55354464-55354464 on Build GRCh38
rs1026270216 Chr.19: 55354402-55354402 on Build GRCh38
rs372120425 Chr.19: 55354459-55354459 on Build GRCh38
rs149294692 Chr.19: 55354427-55354427 on Build GRCh38
rs749659054 Chr.19: 55354380-55354380 on Build GRCh38
rs780482938 Chr.19: 55354374-55354374 on Build GRCh38
rs780160808 Chr.19: 55354500-55354500 on Build GRCh38
rs752994518 Chr.19: 55354490-55354490 on Build GRCh38
rs553344492 Chr.19: 55354329-55354329 on Build GRCh38

View Full Product Details