Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35013333-35013594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1A
Entrez Gene ID
57655
Gene Name
GRAM domain containing 1A
Gene Aliases
KIAA1533
UniGene
Hs.515351
Gene Chromosome Location
Chr.19: 34994784-35026471 on Build GRCh38
Transcripts
NM_001136199.2 NM_001320034.1 NM_001320035.1 NM_001320036.1 NM_020895.4 XM_011527149.1 XM_011527153.1 XM_011527154.2 XM_011527155.1 XM_011527156.1 XM_017027034.1 XM_017027035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6674758 c.748G>A p.A250T Chr.19: 35013569-35013569 on Build GRCh38
4896063 c.764C>T p.T255I Chr.19: 35013585-35013585 on Build GRCh38
6993479 c.685G>A p.D229N Chr.19: 35013334-35013334 on Build GRCh38
4076917 c.747C>T p.I249I Chr.19: 35013568-35013568 on Build GRCh38
6674755 c.752T>C p.L251P Chr.19: 35013573-35013573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186535241 Chr.19: 35013506-35013506 on Build GRCh38
rs866679180 Chr.19: 35013355-35013355 on Build GRCh38
rs761916127 Chr.19: 35013555-35013555 on Build GRCh38
rs147792108 Chr.19: 35013507-35013508 on Build GRCh38
rs113042980 Chr.19: 35013510-35013510 on Build GRCh38
rs773911709 Chr.19: 35013491-35013491 on Build GRCh38
rs372009504 Chr.19: 35013543-35013543 on Build GRCh38
rs778376525 Chr.19: 35013420-35013420 on Build GRCh38
rs373458776 Chr.19: 35013360-35013360 on Build GRCh38
rs200884983 Chr.19: 35013571-35013571 on Build GRCh38
rs533809888 Chr.19: 35013529-35013529 on Build GRCh38
rs774742919 Chr.19: 35013547-35013547 on Build GRCh38
rs762004468 Chr.19: 35013388-35013388 on Build GRCh38
rs747610260 Chr.19: 35013496-35013496 on Build GRCh38
rs568042025 Chr.19: 35013520-35013520 on Build GRCh38
rs767644688 Chr.19: 35013568-35013568 on Build GRCh38
rs985246084 Chr.19: 35013428-35013429 on Build GRCh38
rs771604303 Chr.19: 35013507-35013507 on Build GRCh38
rs568824598 Chr.19: 35013416-35013416 on Build GRCh38
rs1048232910 Chr.19: 35013577-35013577 on Build GRCh38
rs144426841 Chr.19: 35013401-35013401 on Build GRCh38
rs763300905 Chr.19: 35013363-35013363 on Build GRCh38
rs754669024 Chr.19: 35013417-35013417 on Build GRCh38
rs770626095 Chr.19: 35013519-35013519 on Build GRCh38
rs953801898 Chr.19: 35013423-35013423 on Build GRCh38
rs965007146 Chr.19: 35013447-35013447 on Build GRCh38
rs754280652 Chr.19: 35013573-35013573 on Build GRCh38
rs1022238405 Chr.19: 35013405-35013405 on Build GRCh38
rs780521482 Chr.19: 35013409-35013409 on Build GRCh38
rs918479702 Chr.19: 35013546-35013546 on Build GRCh38
rs778409306 Chr.19: 35013394-35013394 on Build GRCh38
rs376334048 Chr.19: 35013379-35013379 on Build GRCh38
rs768094373 Chr.19: 35013389-35013389 on Build GRCh38
rs768950150 Chr.19: 35013533-35013533 on Build GRCh38
rs777505448 Chr.19: 35013509-35013509 on Build GRCh38
rs907248598 Chr.19: 35013362-35013362 on Build GRCh38
rs764237876 Chr.19: 35013375-35013375 on Build GRCh38
rs912222570 Chr.19: 35013446-35013446 on Build GRCh38
rs57653972 Chr.19: 35013510-35013511 on Build GRCh38
rs564732032 Chr.19: 35013469-35013469 on Build GRCh38
rs750877472 Chr.19: 35013569-35013569 on Build GRCh38
rs768168419 Chr.19: 35013470-35013470 on Build GRCh38
rs1002837470 Chr.19: 35013377-35013377 on Build GRCh38

View Full Product Details