Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65330993-65331262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGDCC3
Entrez Gene ID
9543
Gene Name
immunoglobulin superfamily DCC subclass member 3
Gene Aliases
HsT18880, PUNC
UniGene
Hs.128292
Gene Chromosome Location
Chr.15: 65327127-65378040 on Build GRCh38
Transcripts
NM_004884.3 XM_011522241.1 XM_011522243.1 XM_011522244.1 XM_011522245.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7280944 c.1433_1434insA p.S479fs*38 Chr.15: 65331177-65331178 on Build GRCh38
964045 c.1407G>A p.E469E Chr.15: 65331204-65331204 on Build GRCh38
4468157 c.1529C>T p.S510F Chr.15: 65331082-65331082 on Build GRCh38
5456500 c.1397A>G p.D466G Chr.15: 65331214-65331214 on Build GRCh38
4691892 c.1404G>A p.P468P Chr.15: 65331207-65331207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987537415 Chr.15: 65331159-65331159 on Build GRCh38
rs754501933 Chr.15: 65331229-65331229 on Build GRCh38
rs779728815 Chr.15: 65331125-65331125 on Build GRCh38
rs770335929 Chr.15: 65331136-65331136 on Build GRCh38
rs777655118 Chr.15: 65331023-65331023 on Build GRCh38
rs368946941 Chr.15: 65331171-65331171 on Build GRCh38
rs753782123 Chr.15: 65331202-65331202 on Build GRCh38
rs980462357 Chr.15: 65331064-65331064 on Build GRCh38
rs757938214 Chr.15: 65331119-65331119 on Build GRCh38
rs753170706 Chr.15: 65331181-65331181 on Build GRCh38
rs757187982 Chr.15: 65331101-65331101 on Build GRCh38
rs758134594 Chr.15: 65331208-65331208 on Build GRCh38
rs754708920 Chr.15: 65331228-65331228 on Build GRCh38
rs376972102 Chr.15: 65331026-65331026 on Build GRCh38
rs749650556 Chr.15: 65331143-65331143 on Build GRCh38
rs756101597 Chr.15: 65331096-65331096 on Build GRCh38
rs777243001 Chr.15: 65331055-65331055 on Build GRCh38
rs368505415 Chr.15: 65331044-65331044 on Build GRCh38
rs746522726 Chr.15: 65331227-65331227 on Build GRCh38
rs763017935 Chr.15: 65331080-65331080 on Build GRCh38
rs771769341 Chr.15: 65331049-65331049 on Build GRCh38
rs372384897 Chr.15: 65331024-65331024 on Build GRCh38
rs753675254 Chr.15: 65331100-65331100 on Build GRCh38
rs200522013 Chr.15: 65331207-65331207 on Build GRCh38
rs752536685 Chr.15: 65331095-65331095 on Build GRCh38
rs765928620 Chr.15: 65331094-65331095 on Build GRCh38
rs759588889 Chr.15: 65331092-65331092 on Build GRCh38
rs755835681 Chr.15: 65331022-65331022 on Build GRCh38
rs536064808 Chr.15: 65331153-65331153 on Build GRCh38
rs943364145 Chr.15: 65331237-65331237 on Build GRCh38
rs766580979 Chr.15: 65331084-65331084 on Build GRCh38
rs762628478 Chr.15: 65331244-65331244 on Build GRCh38
rs773657976 Chr.15: 65331140-65331140 on Build GRCh38
rs970363087 Chr.15: 65331057-65331057 on Build GRCh38
rs372201182 Chr.15: 65331099-65331099 on Build GRCh38
rs552951892 Chr.15: 65331158-65331158 on Build GRCh38
rs762409532 Chr.15: 65331060-65331060 on Build GRCh38
rs771332776 Chr.15: 65331147-65331147 on Build GRCh38
rs779361046 Chr.15: 65331212-65331212 on Build GRCh38
rs536780229 Chr.15: 65331088-65331088 on Build GRCh38
rs556723208 Chr.15: 65331127-65331127 on Build GRCh38
rs770002061 Chr.15: 65331072-65331072 on Build GRCh38
rs745568465 Chr.15: 65331108-65331108 on Build GRCh38
rs764091980 Chr.15: 65331173-65331173 on Build GRCh38
rs1024623144 Chr.15: 65331036-65331036 on Build GRCh38
rs751192856 Chr.15: 65331159-65331168 on Build GRCh38
rs929633153 Chr.15: 65331071-65331071 on Build GRCh38
rs775654125 Chr.15: 65331168-65331168 on Build GRCh38
rs778699105 Chr.15: 65331104-65331104 on Build GRCh38
rs773636257 Chr.15: 65331077-65331077 on Build GRCh38
rs778503158 Chr.15: 65331027-65331027 on Build GRCh38
rs567089486 Chr.15: 65331152-65331152 on Build GRCh38
rs926518917 Chr.15: 65331074-65331074 on Build GRCh38
rs757100151 Chr.15: 65331204-65331204 on Build GRCh38
rs767625205 Chr.15: 65331094-65331094 on Build GRCh38

View Full Product Details