Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50487466-50487733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIOX
Entrez Gene ID
55586
Gene Name
myo-inositol oxygenase
Gene Aliases
ALDRL6
UniGene
Hs.129227
Gene Chromosome Location
Chr.22: 50483988-50490321 on Build GRCh38
Transcripts
NM_017584.5 XM_005261925.3 XM_011530705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3964437 c.150C>T p.H50H Chr.22: 50487715-50487715 on Build GRCh38
3424273 c.165C>T p.F55F Chr.22: 50487730-50487730 on Build GRCh38
229627 c.132C>G p.Y44* Chr.22: 50487697-50487697 on Build GRCh38
5509273 c.97-1G>T p.? Chr.22: 50487661-50487661 on Build GRCh38
7211187 c.106C>T p.L36F Chr.22: 50487671-50487671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757138171 Chr.22: 50487711-50487716 on Build GRCh38
rs960908091 Chr.22: 50487595-50487595 on Build GRCh38
rs143775710 Chr.22: 50487514-50487514 on Build GRCh38
rs1023734248 Chr.22: 50487603-50487603 on Build GRCh38
rs8136078 Chr.22: 50487586-50487586 on Build GRCh38
rs8140944 Chr.22: 50487638-50487638 on Build GRCh38
rs765172916 Chr.22: 50487655-50487656 on Build GRCh38
rs1049083027 Chr.22: 50487701-50487701 on Build GRCh38
rs759495636 Chr.22: 50487508-50487508 on Build GRCh38
rs992054756 Chr.22: 50487599-50487599 on Build GRCh38
rs370588365 Chr.22: 50487637-50487637 on Build GRCh38
rs796985188 Chr.22: 50487491-50487491 on Build GRCh38
rs763980178 Chr.22: 50487683-50487683 on Build GRCh38
rs951253360 Chr.22: 50487496-50487496 on Build GRCh38
rs764681664 Chr.22: 50487689-50487689 on Build GRCh38
rs190030341 Chr.22: 50487618-50487618 on Build GRCh38
rs35968105 Chr.22: 50487508-50487508 on Build GRCh38
rs1040607009 Chr.22: 50487666-50487666 on Build GRCh38
rs769911163 Chr.22: 50487492-50487492 on Build GRCh38
rs760482854 Chr.22: 50487681-50487681 on Build GRCh38
rs749895311 Chr.22: 50487691-50487691 on Build GRCh38
rs1016749486 Chr.22: 50487708-50487708 on Build GRCh38
rs931997494 Chr.22: 50487685-50487685 on Build GRCh38
rs747313438 Chr.22: 50487639-50487639 on Build GRCh38
rs112424283 Chr.22: 50487555-50487555 on Build GRCh38
rs778475911 Chr.22: 50487627-50487627 on Build GRCh38
rs554830267 Chr.22: 50487711-50487711 on Build GRCh38
rs960982359 Chr.22: 50487580-50487580 on Build GRCh38
rs767460386 Chr.22: 50487513-50487513 on Build GRCh38
rs548846718 Chr.22: 50487537-50487537 on Build GRCh38
rs374038729 Chr.22: 50487652-50487652 on Build GRCh38
rs375908737 Chr.22: 50487619-50487619 on Build GRCh38
rs759577235 Chr.22: 50487677-50487677 on Build GRCh38
rs1016948793 Chr.22: 50487518-50487518 on Build GRCh38
rs375419173 Chr.22: 50487680-50487680 on Build GRCh38
rs761730052 Chr.22: 50487491-50487491 on Build GRCh38
rs149909007 Chr.22: 50487687-50487687 on Build GRCh38
rs2232880 Chr.22: 50487653-50487653 on Build GRCh38
rs773311039 Chr.22: 50487670-50487670 on Build GRCh38
rs969106346 Chr.22: 50487604-50487604 on Build GRCh38
rs78273329 Chr.22: 50487703-50487703 on Build GRCh38
rs952095350 Chr.22: 50487562-50487562 on Build GRCh38
rs201501795 Chr.22: 50487641-50487641 on Build GRCh38
rs781333552 Chr.22: 50487651-50487651 on Build GRCh38
rs1014760912 Chr.22: 50487592-50487592 on Build GRCh38
rs774212211 Chr.22: 50487506-50487506 on Build GRCh38
rs35664871 Chr.22: 50487511-50487512 on Build GRCh38
rs752055088 Chr.22: 50487709-50487709 on Build GRCh38
rs751574612 Chr.22: 50487659-50487686 on Build GRCh38
rs370269290 Chr.22: 50487655-50487655 on Build GRCh38
rs3747273 Chr.22: 50487568-50487568 on Build GRCh38
rs759693673 Chr.22: 50487512-50487514 on Build GRCh38
rs568435187 Chr.22: 50487679-50487679 on Build GRCh38
rs758916346 Chr.22: 50487698-50487698 on Build GRCh38
rs750945165 Chr.22: 50487630-50487630 on Build GRCh38
rs768814045 Chr.22: 50487490-50487490 on Build GRCh38
rs56311845 Chr.22: 50487486-50487486 on Build GRCh38
rs199790962 Chr.22: 50487616-50487616 on Build GRCh38
rs756786668 Chr.22: 50487622-50487622 on Build GRCh38

View Full Product Details