Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33417104-33417375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUTA
Entrez Gene ID
51596
Gene Name
cutA divalent cation tolerance homolog (E. coli)
Gene Aliases
ACHAP, C6orf82
UniGene
Hs.520070
Gene Chromosome Location
Chr.6: 33416542-33418288 on Build GRCh38
Transcripts
NM_001014433.2 NM_001014837.1 NM_001014838.1 NM_001014840.1 NM_015921.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7094752 c.388G>A p.G130R Chr.6: 33417129-33417129 on Build GRCh38
7116897 c.235C>G p.Q79E Chr.6: 33417264-33417264 on Build GRCh38
3625734 c.231C>T p.I77I Chr.6: 33417268-33417268 on Build GRCh38
7094754 c.262G>A p.G88R Chr.6: 33417129-33417129 on Build GRCh38
3674680 c.193G>A p.V65M Chr.6: 33417306-33417306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779722615 Chr.6: 33417228-33417229 on Build GRCh38
rs754996081 Chr.6: 33417298-33417300 on Build GRCh38
rs779034201 Chr.6: 33417324-33417325 on Build GRCh38
rs771789778 Chr.6: 33417151-33417151 on Build GRCh38
rs755582097 Chr.6: 33417160-33417161 on Build GRCh38
rs781056353 Chr.6: 33417315-33417315 on Build GRCh38
rs763754766 Chr.6: 33417157-33417157 on Build GRCh38
rs762047549 Chr.6: 33417216-33417216 on Build GRCh38
rs755377489 Chr.6: 33417261-33417261 on Build GRCh38
rs546082624 Chr.6: 33417349-33417349 on Build GRCh38
rs906720222 Chr.6: 33417288-33417288 on Build GRCh38
rs750106394 Chr.6: 33417153-33417154 on Build GRCh38
rs774447638 Chr.6: 33417336-33417336 on Build GRCh38
rs749384113 Chr.6: 33417179-33417179 on Build GRCh38
rs374469503 Chr.6: 33417310-33417310 on Build GRCh38
rs371639766 Chr.6: 33417275-33417275 on Build GRCh38
rs963586093 Chr.6: 33417344-33417344 on Build GRCh38
rs753791220 Chr.6: 33417293-33417293 on Build GRCh38
rs774357947 Chr.6: 33417208-33417208 on Build GRCh38
rs770813789 Chr.6: 33417160-33417160 on Build GRCh38
rs769571423 Chr.6: 33417333-33417333 on Build GRCh38
rs143728019 Chr.6: 33417313-33417313 on Build GRCh38
rs781310029 Chr.6: 33417341-33417342 on Build GRCh38
rs754368338 Chr.6: 33417242-33417242 on Build GRCh38
rs753779094 Chr.6: 33417229-33417229 on Build GRCh38
rs1021925506 Chr.6: 33417325-33417325 on Build GRCh38
rs752202915 Chr.6: 33417291-33417291 on Build GRCh38
rs773608015 Chr.6: 33417230-33417230 on Build GRCh38
rs1015226219 Chr.6: 33417343-33417343 on Build GRCh38
rs747944386 Chr.6: 33417331-33417334 on Build GRCh38
rs900602187 Chr.6: 33417147-33417147 on Build GRCh38
rs766858212 Chr.6: 33417234-33417234 on Build GRCh38
rs868210240 Chr.6: 33417266-33417266 on Build GRCh38
rs776608716 Chr.6: 33417167-33417167 on Build GRCh38
rs754501395 Chr.6: 33417134-33417134 on Build GRCh38
rs530831916 Chr.6: 33417316-33417316 on Build GRCh38
rs369482910 Chr.6: 33417154-33417154 on Build GRCh38
rs73402303 Chr.6: 33417229-33417229 on Build GRCh38
rs768775576 Chr.6: 33417201-33417201 on Build GRCh38
rs368176232 Chr.6: 33417233-33417233 on Build GRCh38
rs1010489520 Chr.6: 33417317-33417317 on Build GRCh38
rs373777295 Chr.6: 33417193-33417193 on Build GRCh38
rs200376394 Chr.6: 33417299-33417299 on Build GRCh38
rs778499708 Chr.6: 33417149-33417149 on Build GRCh38
rs747809433 Chr.6: 33417150-33417150 on Build GRCh38
rs757061697 Chr.6: 33417314-33417314 on Build GRCh38

View Full Product Details