Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155169259-155169520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTCAP2
Entrez Gene ID
200185
Gene Name
keratinocyte associated protein 2
Gene Aliases
KCP2
UniGene
Hs.516671
Gene Chromosome Location
Chr.1: 155169408-155173328 on Build GRCh38
Transcripts
NM_173852.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896777 c.482G>A p.R161K Chr.1: 155169447-155169447 on Build GRCh38
1334560 c.463A>G p.T155A Chr.1: 155169466-155169466 on Build GRCh38
5479640 c.436G>A p.A146T Chr.1: 155169493-155169493 on Build GRCh38
5353138 c.466G>A p.G156S Chr.1: 155169463-155169463 on Build GRCh38
4494844 c.440C>T p.P147L Chr.1: 155169489-155169489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002747663 Chr.1: 155169325-155169325 on Build GRCh38
rs893794038 Chr.1: 155169312-155169312 on Build GRCh38
rs769159758 Chr.1: 155169372-155169372 on Build GRCh38
rs762559421 Chr.1: 155169469-155169469 on Build GRCh38
rs767417973 Chr.1: 155169487-155169487 on Build GRCh38
rs982903102 Chr.1: 155169455-155169455 on Build GRCh38
rs543580997 Chr.1: 155169311-155169311 on Build GRCh38
rs776182694 Chr.1: 155169381-155169381 on Build GRCh38
rs754757064 Chr.1: 155169485-155169485 on Build GRCh38
rs772774857 Chr.1: 155169443-155169443 on Build GRCh38
rs772295303 Chr.1: 155169337-155169337 on Build GRCh38
rs777035858 Chr.1: 155169376-155169376 on Build GRCh38
rs752593729 Chr.1: 155169489-155169489 on Build GRCh38
rs991437220 Chr.1: 155169412-155169412 on Build GRCh38
rs144875396 Chr.1: 155169381-155169381 on Build GRCh38
rs764639348 Chr.1: 155169438-155169438 on Build GRCh38
rs563765889 Chr.1: 155169291-155169291 on Build GRCh38
rs528947305 Chr.1: 155169499-155169499 on Build GRCh38
rs908331937 Chr.1: 155169478-155169478 on Build GRCh38
rs766068516 Chr.1: 155169473-155169473 on Build GRCh38
rs775968100 Chr.1: 155169361-155169361 on Build GRCh38
rs751303947 Chr.1: 155169480-155169480 on Build GRCh38
rs761282454 Chr.1: 155169411-155169414 on Build GRCh38
rs757269090 Chr.1: 155169498-155169498 on Build GRCh38
rs747427738 Chr.1: 155169369-155169369 on Build GRCh38
rs575773479 Chr.1: 155169308-155169308 on Build GRCh38

View Full Product Details