Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170167429-170167670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL11B
Entrez Gene ID
149281
Gene Name
methyltransferase like 11B
Gene Aliases
C1orf184, HOMT1B, NTM1B
UniGene
Hs.553612
Gene Chromosome Location
Chr.1: 170146047-170168841 on Build GRCh38
Transcripts
NM_001136107.1 XM_011509232.2 XM_011509233.2 XM_011509234.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5551788 c.592G>C p.D198H Chr.1: 170167497-170167497 on Build GRCh38
6541227 c.663G>T p.K221N Chr.1: 170167568-170167568 on Build GRCh38
5906633 c.608C>A p.A203E Chr.1: 170167513-170167513 on Build GRCh38
7187993 c.704A>G p.D235G Chr.1: 170167609-170167609 on Build GRCh38
3771517 c.727G>A p.D243N Chr.1: 170167632-170167632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039916966 Chr.1: 170167480-170167480 on Build GRCh38
rs758956558 Chr.1: 170167522-170167522 on Build GRCh38
rs12735494 Chr.1: 170167644-170167644 on Build GRCh38
rs191834724 Chr.1: 170167538-170167538 on Build GRCh38
rs200874540 Chr.1: 170167514-170167514 on Build GRCh38
rs755280462 Chr.1: 170167499-170167499 on Build GRCh38
rs1023526554 Chr.1: 170167576-170167576 on Build GRCh38
rs199970325 Chr.1: 170167625-170167625 on Build GRCh38
rs936470666 Chr.1: 170167529-170167529 on Build GRCh38
rs373314442 Chr.1: 170167457-170167457 on Build GRCh38
rs12075997 Chr.1: 170167646-170167646 on Build GRCh38
rs996638221 Chr.1: 170167591-170167591 on Build GRCh38
rs778435238 Chr.1: 170167524-170167524 on Build GRCh38
rs768579360 Chr.1: 170167530-170167530 on Build GRCh38
rs564380369 Chr.1: 170167630-170167630 on Build GRCh38
rs78678262 Chr.1: 170167531-170167531 on Build GRCh38
rs887970382 Chr.1: 170167612-170167612 on Build GRCh38
rs750857338 Chr.1: 170167456-170167456 on Build GRCh38
rs554501527 Chr.1: 170167601-170167601 on Build GRCh38
rs779802357 Chr.1: 170167568-170167568 on Build GRCh38
rs773166059 Chr.1: 170167627-170167627 on Build GRCh38
rs943929900 Chr.1: 170167554-170167554 on Build GRCh38
rs748868209 Chr.1: 170167582-170167582 on Build GRCh38
rs866094853 Chr.1: 170167558-170167558 on Build GRCh38
rs934308830 Chr.1: 170167525-170167525 on Build GRCh38
rs779119366 Chr.1: 170167506-170167506 on Build GRCh38
rs917916198 Chr.1: 170167508-170167508 on Build GRCh38
rs1040773979 Chr.1: 170167581-170167581 on Build GRCh38
rs766426941 Chr.1: 170167641-170167641 on Build GRCh38
rs754031826 Chr.1: 170167490-170167490 on Build GRCh38
rs774011868 Chr.1: 170167596-170167596 on Build GRCh38
rs948198277 Chr.1: 170167513-170167513 on Build GRCh38
rs768395103 Chr.1: 170167594-170167594 on Build GRCh38
rs753994106 Chr.1: 170167642-170167642 on Build GRCh38
rs535690075 Chr.1: 170167454-170167454 on Build GRCh38
rs569873347 Chr.1: 170167450-170167450 on Build GRCh38
rs763548960 Chr.1: 170167518-170167518 on Build GRCh38
rs1027121710 Chr.1: 170167608-170167608 on Build GRCh38

View Full Product Details