Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159015795-159016068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI16
Entrez Gene ID
3428
Gene Name
interferon gamma inducible protein 16
Gene Aliases
IFNGIP1, PYHIN2
UniGene
Hs.380250
Gene Chromosome Location
Chr.1: 158999968-159055155 on Build GRCh38
Transcripts
NM_001206567.1 NM_005531.2 XM_005245127.3 XM_006711290.1 XM_017001149.1 XM_017001150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371781 c.277G>A p.A93T Chr.1: 159015883-159015883 on Build GRCh38
7371787 c.310G>A p.A104T Chr.1: 159015916-159015916 on Build GRCh38
7371780 c.277G>A p.A93T Chr.1: 159015883-159015883 on Build GRCh38
4799724 c.275C>A p.P92Q Chr.1: 159015881-159015881 on Build GRCh38
5951697 c.349A>C p.T117P Chr.1: 159015955-159015955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777009420 Chr.1: 159015980-159015980 on Build GRCh38
rs757299472 Chr.1: 159015881-159015881 on Build GRCh38
rs772407332 Chr.1: 159016040-159016040 on Build GRCh38
rs762736524 Chr.1: 159015850-159015850 on Build GRCh38
rs772839461 Chr.1: 159015935-159015935 on Build GRCh38
rs750596025 Chr.1: 159015888-159015888 on Build GRCh38
rs140368446 Chr.1: 159015916-159015916 on Build GRCh38
rs779963703 Chr.1: 159015906-159015906 on Build GRCh38
rs767856907 Chr.1: 159015857-159015857 on Build GRCh38
rs764527673 Chr.1: 159015978-159015978 on Build GRCh38
rs754822963 Chr.1: 159016038-159016038 on Build GRCh38
rs769427556 Chr.1: 159015846-159015846 on Build GRCh38
rs766729623 Chr.1: 159016033-159016033 on Build GRCh38
rs770613302 Chr.1: 159015947-159015947 on Build GRCh38
rs4262539 Chr.1: 159015862-159015862 on Build GRCh38
rs751337977 Chr.1: 159016036-159016036 on Build GRCh38
rs781285207 Chr.1: 159015918-159015918 on Build GRCh38
rs776240321 Chr.1: 159015835-159015835 on Build GRCh38
rs910566363 Chr.1: 159015839-159015839 on Build GRCh38
rs998837305 Chr.1: 159015891-159015891 on Build GRCh38
rs200949538 Chr.1: 159015866-159015866 on Build GRCh38
rs1010663714 Chr.1: 159015962-159015962 on Build GRCh38
rs758694543 Chr.1: 159015896-159015896 on Build GRCh38
rs551655322 Chr.1: 159015925-159015925 on Build GRCh38
rs763960881 Chr.1: 159015965-159015965 on Build GRCh38
rs748873871 Chr.1: 159015942-159015942 on Build GRCh38
rs761050302 Chr.1: 159015973-159015973 on Build GRCh38
rs536691090 Chr.1: 159016042-159016042 on Build GRCh38
rs769180670 Chr.1: 159015929-159015929 on Build GRCh38
rs776418331 Chr.1: 159016028-159016028 on Build GRCh38
rs952163258 Chr.1: 159015819-159015819 on Build GRCh38
rs758653009 Chr.1: 159016024-159016024 on Build GRCh38
rs116270651 Chr.1: 159015883-159015883 on Build GRCh38
rs367582070 Chr.1: 159015969-159015969 on Build GRCh38
rs1017737131 Chr.1: 159016009-159016009 on Build GRCh38
rs374897087 Chr.1: 159015815-159015815 on Build GRCh38
rs1008054467 Chr.1: 159015983-159015983 on Build GRCh38
rs547200732 Chr.1: 159015904-159015904 on Build GRCh38
rs762450597 Chr.1: 159016008-159016008 on Build GRCh38
rs1044726585 Chr.1: 159015976-159015976 on Build GRCh38
rs954756922 Chr.1: 159015943-159015943 on Build GRCh38
rs772740556 Chr.1: 159015849-159015849 on Build GRCh38
rs202135473 Chr.1: 159016011-159016011 on Build GRCh38
rs764617522 Chr.1: 159015877-159015877 on Build GRCh38
rs747629421 Chr.1: 159015841-159015841 on Build GRCh38
rs1057018 Chr.1: 159015913-159015913 on Build GRCh38
rs142497515 Chr.1: 159015878-159015878 on Build GRCh38
rs761142424 Chr.1: 159015868-159015868 on Build GRCh38
rs768754056 Chr.1: 159015832-159015832 on Build GRCh38
rs374626639 Chr.1: 159015914-159015914 on Build GRCh38
rs528236836 Chr.1: 159015958-159015958 on Build GRCh38
rs781105954 Chr.1: 159016039-159016039 on Build GRCh38
rs141289651 Chr.1: 159015931-159015931 on Build GRCh38
rs12566658 Chr.1: 159016028-159016028 on Build GRCh38
rs527906750 Chr.1: 159015829-159015829 on Build GRCh38
rs66519738 Chr.1: 159016041-159016041 on Build GRCh38

View Full Product Details