Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56657666-56657929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOPX
Entrez Gene ID
84525
Gene Name
HOP homeobox
Gene Aliases
CAMEO, HOD, HOP, LAGY, NECC1, OB1, SMAP31, TOTO
UniGene
Hs.619396
Gene Chromosome Location
Chr.4: 56647988-56681866 on Build GRCh38
Transcripts
NM_001145460.1 NM_032495.5 NM_139212.3 XM_006714052.2 XM_017008728.1 XM_017008729.1 XM_017008730.1 XM_017008732.1 XM_017008733.1

SNP Detail

SNP ID Chromosome Location
rs60385558 Chr.4: 56657726-56657726 on Build GRCh38
rs528392952 Chr.4: 56657864-56657864 on Build GRCh38
rs551622354 Chr.4: 56657863-56657863 on Build GRCh38
rs780518020 Chr.4: 56657727-56657727 on Build GRCh38
rs1046801905 Chr.4: 56657893-56657893 on Build GRCh38
rs7693834 Chr.4: 56657703-56657703 on Build GRCh38
rs952747441 Chr.4: 56657800-56657800 on Build GRCh38
rs778299584 Chr.4: 56657801-56657801 on Build GRCh38
rs892507323 Chr.4: 56657859-56657859 on Build GRCh38
rs911344617 Chr.4: 56657815-56657815 on Build GRCh38
rs976912120 Chr.4: 56657837-56657837 on Build GRCh38
rs749858600 Chr.4: 56657750-56657750 on Build GRCh38
rs375212660 Chr.4: 56657879-56657879 on Build GRCh38
rs371825375 Chr.4: 56657839-56657839 on Build GRCh38
rs1053807949 Chr.4: 56657819-56657819 on Build GRCh38
rs368669192 Chr.4: 56657763-56657763 on Build GRCh38
rs1054060400 Chr.4: 56657872-56657872 on Build GRCh38
rs987502274 Chr.4: 56657686-56657686 on Build GRCh38
rs866080230 Chr.4: 56657888-56657888 on Build GRCh38
rs747764961 Chr.4: 56657810-56657810 on Build GRCh38
rs754497084 Chr.4: 56657789-56657789 on Build GRCh38
rs752613234 Chr.4: 56657894-56657894 on Build GRCh38
rs965516381 Chr.4: 56657822-56657822 on Build GRCh38
rs1038869602 Chr.4: 56657730-56657730 on Build GRCh38
rs947079536 Chr.4: 56657899-56657899 on Build GRCh38
rs914182818 Chr.4: 56657878-56657878 on Build GRCh38

View Full Product Details