Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196141154-196141385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK17B
Entrez Gene ID
9262
Gene Name
serine/threonine kinase 17b
Gene Aliases
DRAK2
UniGene
Hs.88297
Gene Chromosome Location
Chr.2: 196133583-196176503 on Build GRCh38
Transcripts
NM_004226.3 XM_011512168.2 XM_011512169.1 XM_011512170.1 XM_011512171.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5963661 c.656+1G>C p.? Chr.2: 196141248-196141248 on Build GRCh38
4503160 c.632C>T p.P211L Chr.2: 196141273-196141273 on Build GRCh38
3575925 c.631C>T p.P211S Chr.2: 196141274-196141274 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376123863 Chr.2: 196141326-196141326 on Build GRCh38
rs781220228 Chr.2: 196141200-196141200 on Build GRCh38
rs1049046818 Chr.2: 196141309-196141309 on Build GRCh38
rs369086051 Chr.2: 196141246-196141246 on Build GRCh38
rs1054457197 Chr.2: 196141189-196141189 on Build GRCh38
rs746242787 Chr.2: 196141209-196141209 on Build GRCh38
rs770632480 Chr.2: 196141276-196141276 on Build GRCh38
rs886463105 Chr.2: 196141348-196141348 on Build GRCh38
rs1010651717 Chr.2: 196141220-196141220 on Build GRCh38
rs765180300 Chr.2: 196141313-196141313 on Build GRCh38
rs769292874 Chr.2: 196141272-196141272 on Build GRCh38
rs756540392 Chr.2: 196141214-196141214 on Build GRCh38
rs749847967 Chr.2: 196141247-196141247 on Build GRCh38
rs763894822 Chr.2: 196141346-196141346 on Build GRCh38
rs772810962 Chr.2: 196141273-196141273 on Build GRCh38
rs1018909007 Chr.2: 196141345-196141345 on Build GRCh38
rs746456183 Chr.2: 196141274-196141274 on Build GRCh38
rs759256129 Chr.2: 196141312-196141312 on Build GRCh38
rs748610659 Chr.2: 196141231-196141234 on Build GRCh38
rs930736864 Chr.2: 196141222-196141222 on Build GRCh38
rs763019792 Chr.2: 196141338-196141338 on Build GRCh38
rs539135098 Chr.2: 196141254-196141254 on Build GRCh38
rs776077952 Chr.2: 196141294-196141294 on Build GRCh38
rs757526546 Chr.2: 196141199-196141199 on Build GRCh38

View Full Product Details