Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158419760-158419997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10K2
Entrez Gene ID
391107
Gene Name
olfactory receptor family 10 subfamily K member 2
Gene Aliases
OR1-4
UniGene
Hs.626815
Gene Chromosome Location
Chr.1: 158419928-158420866 on Build GRCh38
Transcripts
NM_001004476.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
897725 c.923C>T p.T308I Chr.1: 158419944-158419944 on Build GRCh38
529645 c.920G>T p.R307I Chr.1: 158419947-158419947 on Build GRCh38
5559261 c.934T>C p.L312L Chr.1: 158419933-158419933 on Build GRCh38
6058963 c.930C>T p.S310S Chr.1: 158419937-158419937 on Build GRCh38
163100 c.877A>G p.R293G Chr.1: 158419990-158419990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183797168 Chr.1: 158419804-158419804 on Build GRCh38
rs985480386 Chr.1: 158419842-158419842 on Build GRCh38
rs552570656 Chr.1: 158419941-158419941 on Build GRCh38
rs920059780 Chr.1: 158419859-158419859 on Build GRCh38
rs757335791 Chr.1: 158419945-158419945 on Build GRCh38
rs765148428 Chr.1: 158419887-158419887 on Build GRCh38
rs369463112 Chr.1: 158419966-158419966 on Build GRCh38
rs188954264 Chr.1: 158419843-158419843 on Build GRCh38
rs910244126 Chr.1: 158419889-158419889 on Build GRCh38
rs374710976 Chr.1: 158419922-158419922 on Build GRCh38
rs750970731 Chr.1: 158419950-158419950 on Build GRCh38
rs951790503 Chr.1: 158419867-158419867 on Build GRCh38
rs549102774 Chr.1: 158419892-158419892 on Build GRCh38
rs534344458 Chr.1: 158419918-158419918 on Build GRCh38
rs766188066 Chr.1: 158419864-158419864 on Build GRCh38
rs57525598 Chr.1: 158419840-158419840 on Build GRCh38
rs374567639 Chr.1: 158419888-158419888 on Build GRCh38
rs577183472 Chr.1: 158419956-158419956 on Build GRCh38
rs962799949 Chr.1: 158419845-158419845 on Build GRCh38
rs779328165 Chr.1: 158419948-158419948 on Build GRCh38
rs77441663 Chr.1: 158419903-158419903 on Build GRCh38
rs766309362 Chr.1: 158419894-158419894 on Build GRCh38
rs944358625 Chr.1: 158419891-158419891 on Build GRCh38
rs758778919 Chr.1: 158419951-158419951 on Build GRCh38
rs761385855 Chr.1: 158419882-158419882 on Build GRCh38
rs1054223758 Chr.1: 158419921-158419921 on Build GRCh38
rs58605957 Chr.1: 158419869-158419869 on Build GRCh38
rs773434547 Chr.1: 158419813-158419813 on Build GRCh38
rs143743518 Chr.1: 158419967-158419967 on Build GRCh38
rs199749324 Chr.1: 158419868-158419868 on Build GRCh38
rs777978469 Chr.1: 158419957-158419957 on Build GRCh38
rs1028942232 Chr.1: 158419800-158419800 on Build GRCh38
rs1046317099 Chr.1: 158419934-158419934 on Build GRCh38

View Full Product Details