Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74148878-74149143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMM21
Entrez Gene ID
29090
Gene Name
translocase of inner mitochondrial membrane 21
Gene Aliases
C18orf55, HSPC154, TIM21
UniGene
Hs.532835
Gene Chromosome Location
Chr.18: 74148511-74158969 on Build GRCh38
Transcripts
NM_014177.2 XM_005266689.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
708952 c.164G>C p.R55T Chr.18: 74148972-74148972 on Build GRCh38
5227535 c.236G>A p.G79D Chr.18: 74149044-74149044 on Build GRCh38
3527517 c.84C>T p.I28I Chr.18: 74148892-74148892 on Build GRCh38
127472 c.267G>A p.Q89Q Chr.18: 74149075-74149075 on Build GRCh38
4959433 c.175G>A p.G59R Chr.18: 74148983-74148983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751600715 Chr.18: 74148956-74148956 on Build GRCh38
rs569608689 Chr.18: 74149063-74149063 on Build GRCh38
rs867635764 Chr.18: 74149000-74149000 on Build GRCh38
rs767235348 Chr.18: 74148911-74148911 on Build GRCh38
rs139456154 Chr.18: 74149080-74149080 on Build GRCh38
rs3737512 Chr.18: 74149043-74149043 on Build GRCh38
rs755347946 Chr.18: 74148923-74148923 on Build GRCh38
rs772527916 Chr.18: 74149081-74149081 on Build GRCh38
rs185377253 Chr.18: 74149056-74149056 on Build GRCh38
rs752110195 Chr.18: 74149036-74149036 on Build GRCh38
rs763408358 Chr.18: 74149017-74149017 on Build GRCh38
rs748198584 Chr.18: 74148949-74148949 on Build GRCh38
rs776387549 Chr.18: 74149105-74149105 on Build GRCh38
rs749259507 Chr.18: 74149073-74149074 on Build GRCh38
rs760309883 Chr.18: 74149003-74149003 on Build GRCh38
rs1042944862 Chr.18: 74148968-74148968 on Build GRCh38
rs750523747 Chr.18: 74149044-74149044 on Build GRCh38
rs763205947 Chr.18: 74149112-74149112 on Build GRCh38
rs771757138 Chr.18: 74148988-74148988 on Build GRCh38
rs761372442 Chr.18: 74148982-74148982 on Build GRCh38
rs764554238 Chr.18: 74149035-74149035 on Build GRCh38
rs573601238 Chr.18: 74148910-74148910 on Build GRCh38
rs371231552 Chr.18: 74148912-74148912 on Build GRCh38
rs528610432 Chr.18: 74148925-74148925 on Build GRCh38
rs772984727 Chr.18: 74148990-74148990 on Build GRCh38
rs200625916 Chr.18: 74149037-74149037 on Build GRCh38
rs769630155 Chr.18: 74148963-74148963 on Build GRCh38
rs148630515 Chr.18: 74148960-74148960 on Build GRCh38
rs749125285 Chr.18: 74148967-74148967 on Build GRCh38
rs770451974 Chr.18: 74149095-74149095 on Build GRCh38
rs753242199 Chr.18: 74148933-74148933 on Build GRCh38
rs151131593 Chr.18: 74149079-74149079 on Build GRCh38
rs375556503 Chr.18: 74149018-74149018 on Build GRCh38
rs547145109 Chr.18: 74148962-74148962 on Build GRCh38
rs757368880 Chr.18: 74148959-74148959 on Build GRCh38
rs1002907077 Chr.18: 74148975-74148975 on Build GRCh38
rs766057902 Chr.18: 74149006-74149006 on Build GRCh38
rs200022512 Chr.18: 74149052-74149052 on Build GRCh38
rs746933845 Chr.18: 74149071-74149071 on Build GRCh38
rs768426545 Chr.18: 74148976-74148976 on Build GRCh38
rs779650220 Chr.18: 74148966-74148966 on Build GRCh38
rs370815819 Chr.18: 74149060-74149060 on Build GRCh38
rs774457041 Chr.18: 74148980-74148980 on Build GRCh38
rs760245172 Chr.18: 74149093-74149093 on Build GRCh38
rs778276713 Chr.18: 74148942-74148942 on Build GRCh38
rs758762695 Chr.18: 74148940-74148940 on Build GRCh38
rs761428169 Chr.18: 74148906-74148906 on Build GRCh38
rs778190520 Chr.18: 74148951-74148955 on Build GRCh38
rs752957466 Chr.18: 74149008-74149008 on Build GRCh38

View Full Product Details