Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49033562-49033813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFE3
Entrez Gene ID
7030
Gene Name
transcription factor binding to IGHM enhancer 3
Gene Aliases
RCCP2, RCCX1, TFEA, bHLHe33
UniGene
Hs.730740
Gene Chromosome Location
Chr.X: 49028726-49043517 on Build GRCh38
Transcripts
NM_001282142.1 NM_006521.5

SNP Detail

SNP ID Chromosome Location
rs369475777 Chr.X: 49033711-49033711 on Build GRCh38
rs782342097 Chr.X: 49033600-49033600 on Build GRCh38
rs781863465 Chr.X: 49033693-49033693 on Build GRCh38
rs782358989 Chr.X: 49033793-49033793 on Build GRCh38
rs372314452 Chr.X: 49033694-49033694 on Build GRCh38
rs782635252 Chr.X: 49033688-49033688 on Build GRCh38
rs369698146 Chr.X: 49033588-49033588 on Build GRCh38
rs782613018 Chr.X: 49033698-49033698 on Build GRCh38
rs782115411 Chr.X: 49033778-49033778 on Build GRCh38
rs890548874 Chr.X: 49033655-49033655 on Build GRCh38
rs782316450 Chr.X: 49033606-49033606 on Build GRCh38
rs886969467 Chr.X: 49033786-49033786 on Build GRCh38
rs782076934 Chr.X: 49033712-49033712 on Build GRCh38
rs782535970 Chr.X: 49033591-49033591 on Build GRCh38
rs782034465 Chr.X: 49033768-49033768 on Build GRCh38
rs185587819 Chr.X: 49033705-49033705 on Build GRCh38
rs782004627 Chr.X: 49033777-49033777 on Build GRCh38
rs201168885 Chr.X: 49033699-49033699 on Build GRCh38
rs782369351 Chr.X: 49033766-49033766 on Build GRCh38
rs782120251 Chr.X: 49033640-49033640 on Build GRCh38
rs782656388 Chr.X: 49033709-49033709 on Build GRCh38
rs782379337 Chr.X: 49033720-49033720 on Build GRCh38
rs1051885706 Chr.X: 49033638-49033638 on Build GRCh38
rs781963537 Chr.X: 49033612-49033612 on Build GRCh38

View Full Product Details