Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109394320-109394551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKHD1L1
Entrez Gene ID
93035
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1
Gene Aliases
PKHDL1
UniGene
Hs.170128
Gene Chromosome Location
Chr.8: 109362477-109536809 on Build GRCh38
Transcripts
NM_177531.4 XM_011517371.2 XM_017013969.1 XM_017013970.1 XM_017013971.1 XM_017013972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3643947 c.777T>C p.V259V Chr.8: 109394445-109394445 on Build GRCh38
4507519 c.754C>T p.P252S Chr.8: 109394422-109394422 on Build GRCh38
6013238 c.757C>A p.Q253K Chr.8: 109394425-109394425 on Build GRCh38
1663528 c.754C>A p.P252T Chr.8: 109394422-109394422 on Build GRCh38
748806 c.805C>T p.Q269* Chr.8: 109394473-109394473 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373945978 Chr.8: 109394513-109394513 on Build GRCh38
rs577624337 Chr.8: 109394469-109394469 on Build GRCh38
rs775790879 Chr.8: 109394434-109394434 on Build GRCh38
rs73309421 Chr.8: 109394442-109394442 on Build GRCh38
rs752378228 Chr.8: 109394473-109394473 on Build GRCh38
rs377273333 Chr.8: 109394507-109394507 on Build GRCh38
rs901672335 Chr.8: 109394362-109394362 on Build GRCh38
rs931916608 Chr.8: 109394423-109394423 on Build GRCh38
rs140463251 Chr.8: 109394517-109394517 on Build GRCh38
rs756332399 Chr.8: 109394464-109394464 on Build GRCh38
rs759770495 Chr.8: 109394403-109394403 on Build GRCh38
rs769935513 Chr.8: 109394516-109394516 on Build GRCh38
rs374287065 Chr.8: 109394490-109394490 on Build GRCh38
rs745875853 Chr.8: 109394514-109394514 on Build GRCh38
rs113740078 Chr.8: 109394368-109394368 on Build GRCh38
rs200518130 Chr.8: 109394427-109394427 on Build GRCh38
rs753221352 Chr.8: 109394467-109394467 on Build GRCh38
rs530313710 Chr.8: 109394452-109394452 on Build GRCh38
rs73309424 Chr.8: 109394448-109394448 on Build GRCh38
rs370573554 Chr.8: 109394509-109394509 on Build GRCh38
rs1023660033 Chr.8: 109394493-109394493 on Build GRCh38
rs77766145 Chr.8: 109394468-109394468 on Build GRCh38
rs746029541 Chr.8: 109394377-109394377 on Build GRCh38
rs570125235 Chr.8: 109394392-109394392 on Build GRCh38
rs1015941466 Chr.8: 109394433-109394433 on Build GRCh38
rs77730643 Chr.8: 109394466-109394466 on Build GRCh38
rs921570411 Chr.8: 109394380-109394380 on Build GRCh38
rs769837952 Chr.8: 109394383-109394383 on Build GRCh38
rs765520503 Chr.8: 109394513-109394514 on Build GRCh38
rs200009645 Chr.8: 109394520-109394520 on Build GRCh38
rs539118717 Chr.8: 109394363-109394363 on Build GRCh38
rs570463070 Chr.8: 109394362-109394362 on Build GRCh38
rs543517137 Chr.8: 109394481-109394481 on Build GRCh38
rs528830726 Chr.8: 109394456-109394456 on Build GRCh38
rs889047988 Chr.8: 109394400-109394400 on Build GRCh38
rs893896613 Chr.8: 109394485-109394485 on Build GRCh38
rs754124635 Chr.8: 109394402-109394403 on Build GRCh38
rs1010968246 Chr.8: 109394486-109394486 on Build GRCh38

View Full Product Details