Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39013870-39014135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM9
Entrez Gene ID
8754
Gene Name
ADAM metallopeptidase domain 9
Gene Aliases
CORD9, MCMP, MDC9, Mltng
UniGene
Hs.591852
Gene Chromosome Location
Chr.8: 38996771-39105261 on Build GRCh38
Transcripts
NM_003816.2 NR_027638.1 NR_027639.1 NR_027878.1 XM_011544682.2 XM_017013942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4779835 c.257A>G p.D86G Chr.8: 39013967-39013967 on Build GRCh38
750434 c.312C>G p.I104M Chr.8: 39014022-39014022 on Build GRCh38
4659411 c.260delT p.L88fs*16 Chr.8: 39013970-39013970 on Build GRCh38
6593750 c.255-1G>A p.? Chr.8: 39013964-39013964 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs550158378 Chr.8: 39014036-39014036 on Build GRCh38
rs771587211 Chr.8: 39014025-39014025 on Build GRCh38
rs759627385 Chr.8: 39014079-39014079 on Build GRCh38
rs138058399 Chr.8: 39014026-39014026 on Build GRCh38
rs752599940 Chr.8: 39013946-39013946 on Build GRCh38
rs372386835 Chr.8: 39014069-39014069 on Build GRCh38
rs77781038 Chr.8: 39013918-39013918 on Build GRCh38
rs540187906 Chr.8: 39013929-39013929 on Build GRCh38
rs570001592 Chr.8: 39014111-39014111 on Build GRCh38
rs766421346 Chr.8: 39013963-39013963 on Build GRCh38
rs970046146 Chr.8: 39013927-39013927 on Build GRCh38
rs777287822 Chr.8: 39014033-39014033 on Build GRCh38
rs561769853 Chr.8: 39014068-39014068 on Build GRCh38
rs769111658 Chr.8: 39013934-39013934 on Build GRCh38
rs886514573 Chr.8: 39013899-39013899 on Build GRCh38
rs773464514 Chr.8: 39014049-39014049 on Build GRCh38
rs533862332 Chr.8: 39013907-39013907 on Build GRCh38
rs75029712 Chr.8: 39013919-39013919 on Build GRCh38
rs191016085 Chr.8: 39013942-39013942 on Build GRCh38
rs770307880 Chr.8: 39014038-39014038 on Build GRCh38
rs756115024 Chr.8: 39013969-39013969 on Build GRCh38
rs745320048 Chr.8: 39013922-39013922 on Build GRCh38
rs763954582 Chr.8: 39014089-39014089 on Build GRCh38
rs777199628 Chr.8: 39013941-39013941 on Build GRCh38
rs776015736 Chr.8: 39013928-39013928 on Build GRCh38
rs745528352 Chr.8: 39014016-39014016 on Build GRCh38
rs752653261 Chr.8: 39013965-39013965 on Build GRCh38
rs763041780 Chr.8: 39013944-39013944 on Build GRCh38
rs201485493 Chr.8: 39014003-39014003 on Build GRCh38
rs747513455 Chr.8: 39014028-39014028 on Build GRCh38
rs757052065 Chr.8: 39014002-39014002 on Build GRCh38
rs753572901 Chr.8: 39014001-39014001 on Build GRCh38
rs76099010 Chr.8: 39013943-39013943 on Build GRCh38
rs146980702 Chr.8: 39013990-39013990 on Build GRCh38
rs773303455 Chr.8: 39013948-39013948 on Build GRCh38
rs771512707 Chr.8: 39013925-39013925 on Build GRCh38
rs763128480 Chr.8: 39014063-39014063 on Build GRCh38
rs1006287529 Chr.8: 39013906-39013906 on Build GRCh38
rs1047540964 Chr.8: 39014113-39014113 on Build GRCh38
rs769340668 Chr.8: 39014032-39014032 on Build GRCh38

View Full Product Details