Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44172985-44173246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN11
Entrez Gene ID
11131
Gene Name
calpain 11
Gene Aliases
calpain11
UniGene
Hs.225953
Gene Chromosome Location
Chr.6: 44158811-44184402 on Build GRCh38
Transcripts
NM_007058.3 XM_006714984.1 XM_006714985.1 XM_006714987.1 XM_011514274.1 XM_011514275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153430 c.662+6T>G p.? Chr.6: 44173079-44173079 on Build GRCh38
3765283 c.662+8C>T p.? Chr.6: 44173081-44173081 on Build GRCh38
150098 c.662+8C>G p.? Chr.6: 44173081-44173081 on Build GRCh38
7359416 c.619C>T p.R207C Chr.6: 44173030-44173030 on Build GRCh38
7229048 c.662+8C>G p.? Chr.6: 44173081-44173081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767123649 Chr.6: 44173054-44173054 on Build GRCh38
rs933872671 Chr.6: 44173183-44173183 on Build GRCh38
rs3813500 Chr.6: 44173081-44173081 on Build GRCh38
rs758767166 Chr.6: 44173082-44173082 on Build GRCh38
rs372617735 Chr.6: 44173031-44173031 on Build GRCh38
rs374999083 Chr.6: 44173013-44173013 on Build GRCh38
rs745954774 Chr.6: 44173169-44173169 on Build GRCh38
rs769594113 Chr.6: 44173020-44173020 on Build GRCh38
rs979545700 Chr.6: 44173118-44173118 on Build GRCh38
rs1029970071 Chr.6: 44173027-44173027 on Build GRCh38
rs938136070 Chr.6: 44173142-44173142 on Build GRCh38
rs775445812 Chr.6: 44173026-44173026 on Build GRCh38
rs200279946 Chr.6: 44173030-44173030 on Build GRCh38
rs771109804 Chr.6: 44173092-44173092 on Build GRCh38
rs769874215 Chr.6: 44173177-44173177 on Build GRCh38
rs371227294 Chr.6: 44173200-44173200 on Build GRCh38
rs774285384 Chr.6: 44173222-44173222 on Build GRCh38
rs751274577 Chr.6: 44173038-44173038 on Build GRCh38
rs779311925 Chr.6: 44173065-44173065 on Build GRCh38
rs768635315 Chr.6: 44173219-44173219 on Build GRCh38
rs918133682 Chr.6: 44173161-44173161 on Build GRCh38
rs992355441 Chr.6: 44173159-44173159 on Build GRCh38
rs747297049 Chr.6: 44173087-44173087 on Build GRCh38
rs749183714 Chr.6: 44173212-44173212 on Build GRCh38
rs566636586 Chr.6: 44173084-44173084 on Build GRCh38
rs534113991 Chr.6: 44173097-44173097 on Build GRCh38
rs985261959 Chr.6: 44173046-44173046 on Build GRCh38
rs776567421 Chr.6: 44173009-44173009 on Build GRCh38
rs929587941 Chr.6: 44173169-44173169 on Build GRCh38
rs761548409 Chr.6: 44173051-44173051 on Build GRCh38
rs373882901 Chr.6: 44173215-44173215 on Build GRCh38
rs886811693 Chr.6: 44173206-44173206 on Build GRCh38
rs761601477 Chr.6: 44173224-44173224 on Build GRCh38
rs755559441 Chr.6: 44173064-44173064 on Build GRCh38
rs552423781 Chr.6: 44173135-44173135 on Build GRCh38
rs749873430 Chr.6: 44173060-44173060 on Build GRCh38

View Full Product Details