Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47590201-47590430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF4
Entrez Gene ID
114900
Gene Name
C1q and tumor necrosis factor related protein 4
Gene Aliases
CTRP4, ZACRP4
UniGene
Hs.662633
Gene Chromosome Location
Chr.11: 47589664-47596336 on Build GRCh38
Transcripts
NM_031909.2 XM_017017165.1 XM_017017166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588824 c.551C>T p.T184M Chr.11: 47590260-47590260 on Build GRCh38
4413160 c.405G>T p.R135R Chr.11: 47590406-47590406 on Build GRCh38
6567782 c.526C>T p.R176C Chr.11: 47590285-47590285 on Build GRCh38
429073 c.588G>A p.P196P Chr.11: 47590223-47590223 on Build GRCh38
5460271 c.534C>T p.A178A Chr.11: 47590277-47590277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904371806 Chr.11: 47590366-47590366 on Build GRCh38
rs1034107198 Chr.11: 47590379-47590379 on Build GRCh38
rs545809872 Chr.11: 47590384-47590384 on Build GRCh38
rs183612270 Chr.11: 47590375-47590375 on Build GRCh38
rs201749016 Chr.11: 47590239-47590239 on Build GRCh38
rs750923662 Chr.11: 47590271-47590271 on Build GRCh38
rs1010055143 Chr.11: 47590342-47590342 on Build GRCh38
rs142453721 Chr.11: 47590388-47590388 on Build GRCh38
rs1015109086 Chr.11: 47590387-47590387 on Build GRCh38
rs968896569 Chr.11: 47590266-47590266 on Build GRCh38
rs777644897 Chr.11: 47590234-47590234 on Build GRCh38
rs892757307 Chr.11: 47590297-47590311 on Build GRCh38
rs751180497 Chr.11: 47590322-47590322 on Build GRCh38
rs770728499 Chr.11: 47590238-47590238 on Build GRCh38
rs567967350 Chr.11: 47590381-47590381 on Build GRCh38
rs1050541034 Chr.11: 47590325-47590325 on Build GRCh38
rs191830241 Chr.11: 47590288-47590288 on Build GRCh38
rs769677404 Chr.11: 47590230-47590230 on Build GRCh38
rs911841118 Chr.11: 47590329-47590329 on Build GRCh38
rs768872465 Chr.11: 47590241-47590241 on Build GRCh38
rs766747599 Chr.11: 47590276-47590276 on Build GRCh38
rs920788819 Chr.11: 47590318-47590318 on Build GRCh38
rs765792933 Chr.11: 47590267-47590267 on Build GRCh38
rs959742864 Chr.11: 47590285-47590285 on Build GRCh38
rs944728116 Chr.11: 47590338-47590338 on Build GRCh38
rs1040975912 Chr.11: 47590339-47590339 on Build GRCh38
rs751216428 Chr.11: 47590224-47590224 on Build GRCh38
rs752151334 Chr.11: 47590382-47590382 on Build GRCh38
rs777611706 Chr.11: 47590392-47590392 on Build GRCh38
rs757058499 Chr.11: 47590399-47590399 on Build GRCh38
rs546482481 Chr.11: 47590389-47590389 on Build GRCh38
rs891633183 Chr.11: 47590233-47590233 on Build GRCh38
rs762443705 Chr.11: 47590258-47590258 on Build GRCh38
rs780620024 Chr.11: 47590227-47590227 on Build GRCh38
rs973563841 Chr.11: 47590310-47590310 on Build GRCh38
rs763271114 Chr.11: 47590272-47590272 on Build GRCh38
rs959690286 Chr.11: 47590380-47590380 on Build GRCh38
rs113439870 Chr.11: 47590354-47590354 on Build GRCh38
rs1034403981 Chr.11: 47590277-47590277 on Build GRCh38
rs748080430 Chr.11: 47590228-47590228 on Build GRCh38
rs1043259089 Chr.11: 47590352-47590352 on Build GRCh38
rs868144458 Chr.11: 47590222-47590222 on Build GRCh38
rs908166662 Chr.11: 47590291-47590292 on Build GRCh38
rs749245388 Chr.11: 47590393-47590393 on Build GRCh38
rs776786694 Chr.11: 47590249-47590249 on Build GRCh38
rs932030824 Chr.11: 47590325-47590326 on Build GRCh38
rs973639114 Chr.11: 47590397-47590397 on Build GRCh38
rs867117652 Chr.11: 47590362-47590362 on Build GRCh38

View Full Product Details