Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 113460068-113460335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
UniGene
Hs.144114
Gene Chromosome Location
Chr.4: 113451032-113761927 on Build GRCh38
Transcripts
NM_001221.3 NM_001321566.1 NM_001321567.1 NM_001321568.1 NM_001321569.1 NM_001321570.1 NM_001321571.1 NM_001321572.1 NM_001321573.1 NM_001321574.1 NM_001321575.1 NM_001321576.1 NM_001321577.1 NM_001321578.1 NM_001321579.1 NM_001321580.1 NM_001321581.1 NM_001321582.1 NM_001321583.1 NM_001321584.1 NM_001321585.1 NM_001321586.1 NM_001321587.1 NM_001321588.1 NM_001321589.1 NM_001321590.1 NM_001321591.1 NM_172114.1 NM_172115.2 NM_172127.2 NM_172128.2 NM_172129.1 XM_005263253.3 XM_005263254.3 XM_006714330.2 XM_011532289.1 XM_011532290.1 XM_011532291.1 XM_011532292.2 XM_011532295.1 XM_017008672.1 XM_017008673.1 XM_017008674.1 XM_017008675.1 XM_017008676.1 XM_017008677.1 XM_017008678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344068 c.1183C>A p.H395N Chr.4: 113460168-113460168 on Build GRCh38
7359338 c.1160G>A p.C387Y Chr.4: 113460233-113460233 on Build GRCh38
7359339 c.1151G>A p.C384Y Chr.4: 113460233-113460233 on Build GRCh38
4329015 c.1158C>A p.I386I Chr.4: 113460235-113460235 on Build GRCh38
6382701 c.1111A>G p.K371E Chr.4: 113460240-113460240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772057103 Chr.4: 113460251-113460251 on Build GRCh38
rs377141236 Chr.4: 113460279-113460279 on Build GRCh38
rs762098380 Chr.4: 113460282-113460282 on Build GRCh38
rs954913640 Chr.4: 113460106-113460106 on Build GRCh38
rs775220710 Chr.4: 113460259-113460259 on Build GRCh38
rs748916709 Chr.4: 113460165-113460165 on Build GRCh38
rs781154752 Chr.4: 113460121-113460123 on Build GRCh38
rs550751815 Chr.4: 113460287-113460287 on Build GRCh38
rs747709397 Chr.4: 113460120-113460120 on Build GRCh38
rs987657983 Chr.4: 113460164-113460164 on Build GRCh38
rs1027727243 Chr.4: 113460293-113460293 on Build GRCh38
rs774126605 Chr.4: 113460223-113460223 on Build GRCh38
rs780627146 Chr.4: 113460107-113460107 on Build GRCh38
rs113419560 Chr.4: 113460268-113460268 on Build GRCh38
rs529121014 Chr.4: 113460142-113460142 on Build GRCh38
rs966459901 Chr.4: 113460278-113460278 on Build GRCh38
rs764035835 Chr.4: 113460272-113460272 on Build GRCh38
rs772949010 Chr.4: 113460155-113460155 on Build GRCh38
rs773115007 Chr.4: 113460108-113460108 on Build GRCh38
rs769119552 Chr.4: 113460122-113460122 on Build GRCh38
rs139199077 Chr.4: 113460224-113460224 on Build GRCh38
rs748039833 Chr.4: 113460280-113460280 on Build GRCh38
rs770752204 Chr.4: 113460175-113460175 on Build GRCh38
rs760837014 Chr.4: 113460262-113460262 on Build GRCh38

View Full Product Details