Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 176884264-176884537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
UniGene
Hs.495897
Gene Chromosome Location
Chr.1: 176855118-177164904 on Build GRCh38
Transcripts
NM_001286164.1 NM_004319.2 NM_207108.2 XM_017001340.1 XM_017001341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4850081 c.3190G>C p.D1064H Chr.1: 176884399-176884399 on Build GRCh38
5435241 c.3184G>A p.E1062K Chr.1: 176884381-176884381 on Build GRCh38
6606146 c.3179G>A p.R1060H Chr.1: 176884386-176884386 on Build GRCh38
5435242 c.3208G>A p.E1070K Chr.1: 176884381-176884381 on Build GRCh38
4850080 c.3166G>C p.D1056H Chr.1: 176884399-176884399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759832557 Chr.1: 176884412-176884412 on Build GRCh38
rs997020667 Chr.1: 176884334-176884334 on Build GRCh38
rs201593312 Chr.1: 176884386-176884386 on Build GRCh38
rs368454490 Chr.1: 176884299-176884299 on Build GRCh38
rs764654112 Chr.1: 176884426-176884426 on Build GRCh38
rs776507266 Chr.1: 176884410-176884410 on Build GRCh38
rs754524217 Chr.1: 176884361-176884361 on Build GRCh38
rs771146109 Chr.1: 176884409-176884409 on Build GRCh38
rs781354760 Chr.1: 176884475-176884475 on Build GRCh38
rs749107910 Chr.1: 176884295-176884295 on Build GRCh38
rs144434019 Chr.1: 176884469-176884469 on Build GRCh38
rs758252814 Chr.1: 176884468-176884468 on Build GRCh38
rs752963618 Chr.1: 176884341-176884341 on Build GRCh38
rs143440206 Chr.1: 176884476-176884476 on Build GRCh38
rs768487237 Chr.1: 176884296-176884296 on Build GRCh38
rs752050989 Chr.1: 176884432-176884432 on Build GRCh38
rs763197786 Chr.1: 176884424-176884424 on Build GRCh38
rs367616283 Chr.1: 176884414-176884414 on Build GRCh38
rs779650754 Chr.1: 176884290-176884290 on Build GRCh38
rs374038452 Chr.1: 176884348-176884348 on Build GRCh38
rs747141288 Chr.1: 176884393-176884393 on Build GRCh38
rs767644396 Chr.1: 176884317-176884317 on Build GRCh38
rs762633579 Chr.1: 176884324-176884325 on Build GRCh38
rs775572710 Chr.1: 176884494-176884494 on Build GRCh38
rs757706796 Chr.1: 176884434-176884434 on Build GRCh38
rs201898410 Chr.1: 176884435-176884435 on Build GRCh38
rs765882689 Chr.1: 176884441-176884441 on Build GRCh38
rs747869835 Chr.1: 176884381-176884381 on Build GRCh38
rs914406225 Chr.1: 176884379-176884379 on Build GRCh38
rs898122560 Chr.1: 176884320-176884320 on Build GRCh38
rs778395608 Chr.1: 176884458-176884458 on Build GRCh38
rs776978957 Chr.1: 176884328-176884328 on Build GRCh38
rs111487131 Chr.1: 176884340-176884340 on Build GRCh38
rs756904416 Chr.1: 176884501-176884501 on Build GRCh38
rs773275290 Chr.1: 176884387-176884387 on Build GRCh38
rs150457018 Chr.1: 176884415-176884415 on Build GRCh38
rs755617903 Chr.1: 176884357-176884357 on Build GRCh38
rs755813890 Chr.1: 176884289-176884289 on Build GRCh38
rs370700052 Chr.1: 176884312-176884312 on Build GRCh38
rs763368936 Chr.1: 176884504-176884504 on Build GRCh38
rs61820881 Chr.1: 176884293-176884293 on Build GRCh38
rs545428774 Chr.1: 176884473-176884473 on Build GRCh38
rs143925375 Chr.1: 176884344-176884344 on Build GRCh38
rs79630456 Chr.1: 176884336-176884336 on Build GRCh38
rs748124885 Chr.1: 176884307-176884307 on Build GRCh38
rs753251844 Chr.1: 176884452-176884452 on Build GRCh38
rs770096764 Chr.1: 176884487-176884487 on Build GRCh38
rs558363072 Chr.1: 176884351-176884351 on Build GRCh38
rs766772171 Chr.1: 176884325-176884325 on Build GRCh38
rs770951753 Chr.1: 176884474-176884474 on Build GRCh38
rs754705091 Chr.1: 176884454-176884454 on Build GRCh38
rs939943483 Chr.1: 176884493-176884494 on Build GRCh38
rs200200255 Chr.1: 176884359-176884359 on Build GRCh38
rs752600572 Chr.1: 176884462-176884462 on Build GRCh38

View Full Product Details