Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 83344817-83345044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH13
Entrez Gene ID
1012
Gene Name
cadherin 13
Gene Aliases
CDHH, P105
UniGene
Hs.654386
Gene Chromosome Location
Chr.16: 82626794-83796610 on Build GRCh38
Transcripts
NM_001220488.1 NM_001220489.1 NM_001220490.1 NM_001257.4 XM_011522804.2 XM_017022848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5368300 c.455G>C p.G152A Chr.16: 83344995-83344995 on Build GRCh38
5200645 c.366G>C p.G122G Chr.16: 83344906-83344906 on Build GRCh38
6624676 c.445G>A p.V149I Chr.16: 83344985-83344985 on Build GRCh38
7282520 c.421C>T p.R141W Chr.16: 83344961-83344961 on Build GRCh38
7103345 c.451G>A p.E151K Chr.16: 83344991-83344991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201452763 Chr.16: 83344909-83344909 on Build GRCh38
rs763794284 Chr.16: 83345013-83345013 on Build GRCh38
rs757067208 Chr.16: 83344981-83344981 on Build GRCh38
rs933818301 Chr.16: 83344971-83344971 on Build GRCh38
rs765363332 Chr.16: 83345000-83345000 on Build GRCh38
rs773529566 Chr.16: 83344908-83344908 on Build GRCh38
rs766559854 Chr.16: 83344916-83344916 on Build GRCh38
rs757167252 Chr.16: 83345018-83345018 on Build GRCh38
rs374048975 Chr.16: 83344887-83344887 on Build GRCh38
rs746970851 Chr.16: 83344878-83344878 on Build GRCh38
rs774758244 Chr.16: 83344900-83344900 on Build GRCh38
rs367662792 Chr.16: 83344984-83344984 on Build GRCh38
rs920619557 Chr.16: 83345016-83345016 on Build GRCh38
rs559519072 Chr.16: 83345014-83345014 on Build GRCh38
rs765328287 Chr.16: 83344925-83344925 on Build GRCh38
rs745434540 Chr.16: 83344968-83344968 on Build GRCh38
rs772306174 Chr.16: 83344979-83344979 on Build GRCh38
rs371020017 Chr.16: 83344976-83344976 on Build GRCh38
rs145120824 Chr.16: 83344882-83344882 on Build GRCh38
rs752994642 Chr.16: 83344926-83344926 on Build GRCh38
rs189965190 Chr.16: 83345010-83345010 on Build GRCh38
rs757253202 Chr.16: 83344954-83344954 on Build GRCh38
rs200608482 Chr.16: 83344862-83344862 on Build GRCh38
rs771613971 Chr.16: 83344985-83344985 on Build GRCh38
rs370508034 Chr.16: 83344987-83344987 on Build GRCh38
rs772486096 Chr.16: 83344901-83344901 on Build GRCh38
rs946751736 Chr.16: 83344876-83344876 on Build GRCh38
rs1002497848 Chr.16: 83345015-83345015 on Build GRCh38
rs369252150 Chr.16: 83344897-83344897 on Build GRCh38
rs1023178211 Chr.16: 83344858-83344858 on Build GRCh38
rs377210458 Chr.16: 83344961-83344961 on Build GRCh38
rs778817843 Chr.16: 83344853-83344853 on Build GRCh38
rs200305692 Chr.16: 83344915-83344919 on Build GRCh38
rs764024071 Chr.16: 83344857-83344857 on Build GRCh38
rs767396298 Chr.16: 83345020-83345020 on Build GRCh38
rs779537085 Chr.16: 83344977-83344977 on Build GRCh38
rs780740509 Chr.16: 83344884-83344884 on Build GRCh38
rs751441653 Chr.16: 83344949-83344949 on Build GRCh38
rs1039625191 Chr.16: 83344931-83344931 on Build GRCh38
rs376338751 Chr.16: 83344970-83344970 on Build GRCh38
rs758063450 Chr.16: 83344927-83344927 on Build GRCh38
rs773691134 Chr.16: 83344982-83344982 on Build GRCh38
rs759648548 Chr.16: 83344990-83344990 on Build GRCh38
rs561374965 Chr.16: 83344870-83344870 on Build GRCh38
rs563977188 Chr.16: 83344924-83344924 on Build GRCh38
rs375664631 Chr.16: 83344978-83344978 on Build GRCh38

View Full Product Details