Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68868304-68868535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA8
Entrez Gene ID
10351
Gene Name
ATP binding cassette subfamily A member 8
Gene Aliases
-
UniGene
Hs.58351
Gene Chromosome Location
Chr.17: 68867285-68955426 on Build GRCh38
Transcripts
NM_001288985.1 NM_001288986.1 NM_007168.3 XM_005256938.2 XM_005256940.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913833 c.4735G>A p.E1579K Chr.17: 68868333-68868333 on Build GRCh38
278518 c.4607A>C p.D1536A Chr.17: 68868341-68868341 on Build GRCh38
3388111 c.4611A>G p.L1537L Chr.17: 68868337-68868337 on Build GRCh38
4068990 c.4742G>A p.S1581N Chr.17: 68868326-68868326 on Build GRCh38
4068989 c.4622G>A p.S1541N Chr.17: 68868326-68868326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768238319 Chr.17: 68868384-68868384 on Build GRCh38
rs564880838 Chr.17: 68868326-68868326 on Build GRCh38
rs576937819 Chr.17: 68868443-68868443 on Build GRCh38
rs960839336 Chr.17: 68868329-68868329 on Build GRCh38
rs967764031 Chr.17: 68868347-68868347 on Build GRCh38
rs576803172 Chr.17: 68868368-68868368 on Build GRCh38
rs755781896 Chr.17: 68868331-68868331 on Build GRCh38
rs747253015 Chr.17: 68868406-68868406 on Build GRCh38
rs756684354 Chr.17: 68868369-68868369 on Build GRCh38
rs541115380 Chr.17: 68868391-68868391 on Build GRCh38
rs758767083 Chr.17: 68868399-68868399 on Build GRCh38
rs367716179 Chr.17: 68868376-68868376 on Build GRCh38
rs776103228 Chr.17: 68868397-68868398 on Build GRCh38
rs371198020 Chr.17: 68868386-68868386 on Build GRCh38
rs763564526 Chr.17: 68868351-68868351 on Build GRCh38
rs530189043 Chr.17: 68868444-68868444 on Build GRCh38
rs891946723 Chr.17: 68868494-68868494 on Build GRCh38
rs766293627 Chr.17: 68868424-68868424 on Build GRCh38
rs980032817 Chr.17: 68868385-68868385 on Build GRCh38
rs1053320897 Chr.17: 68868468-68868468 on Build GRCh38
rs779282773 Chr.17: 68868421-68868421 on Build GRCh38
rs548386936 Chr.17: 68868447-68868447 on Build GRCh38
rs780393199 Chr.17: 68868404-68868404 on Build GRCh38
rs867662408 Chr.17: 68868478-68868478 on Build GRCh38

View Full Product Details